Learn More
Haldenwanger™ Porcelain Spouted Mortar
Use these mortars for a variety of laboratory applications. Haldenwanger™ Porcelain Spouted Mortars are constructed from optimized laboratory porcelain.
Brand:  Haldenwanger™ 55/5/glazed
 View more versions of this product
Product Code. 10720603
  • £125.00 / Each
Estimated Shipment: 15-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Available in a variety of sizes
  • With spout for easy pouring
  • Complies with DIN VDE 0335, type C 100
Specifications
Specifications
400 mL
Mortar
With spout
130mm
Porcelain
Mortar