Learn More
Haldenwanger™ Porcelain Spouted Mortar
Use these mortars for a variety of laboratory applications. Haldenwanger™ Porcelain Spouted Mortars are constructed from optimized laboratory porcelain.
Brand:  Haldenwanger™ 55/6A
 View more versions of this product
Product Code. 12363328
  • £245.00 / Each
Estimated Shipment: 15-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Available in a variety of sizes
  • With spout for easy pouring
  • Complies with DIN VDE 0335, type C 100
Specifications
Specifications
650 mL
Mortar
70mm
Mortar
150mm
Porcelain
With spout
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products