Learn More
Haldenwanger™ Porcelain Spouted Mortar
Use these mortars for a variety of laboratory applications. Haldenwanger™ Porcelain Spouted Mortars are constructed from optimized laboratory porcelain.
Brand:  Haldenwanger™ 55/8
 View more versions of this product
Product Code. 12373328
  • £360.00 / Each
Estimated Shipment: 06-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Grinding surfaces are unglazed.
Specifications
Specifications
1000 mL
Mortar
80mm
Mortar
180mm
Porcelain
With spout
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products