Learn More
Thermo Scientific™ POROS™ CaptureSelect™ AAVX Affinity Resin
POROS CaptureSelect AAVX Affinity Resin is engineered to address the high selectivity and capacity requirements for large-scale downstream purification of a broad range of naturally occurring and synthetic adeno-associated virus (AAV) serotypes used for gene therapy applications.
£1428.00 - £6290.00
Specifications
Binding Capacity | >1 x 1013 genome copies/mL of resin |
---|---|
Content And Storage | Store in refrigerator at 2°C to 8°C. |
Shipping Condition | Wet Ice |
Particle Size | 50 μm |
Form | Liquid Suspension |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18486528
|
Thermo Scientific™
A36739 |
10 mL |
£1428.00
10mL |
Estimated Shipment: 09-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
18406688
|
Thermo Scientific™
A36741 |
50 mL |
£6290.00
50mL |
Estimated Shipment: 09-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
POROS CaptureSelect AAVX Affinity Resin is engineered to address the high selectivity and capacity requirements for large-scale downstream purification of a broad range of naturally occurring and synthetic adeno-associated virus (AAV) serotypes used for gene therapy applications. POROS CaptureSelect AAVX resin has demonstrated binding reactivity towards a set of AAV serotypes that includes AAV1 to AAV8, and AAVrh10 as well as synthetic serotypes.
Features of POROS CaptureSelect AAVX resin include:
- One-step AAV purification from crude material with high purity and yield
- High specificity and capacity, maximizing yield by significantly reducing process steps
- Superior capacity maintained at high flow rates
- Excellent scalability and easier handling and packing due to rigid polymeric bead
- Non-animal derived
POROS CaptureSelect AAVX Affinity Resin enables a robust, fast, and platformable purification process for wide-ranging AAV serotypes with excellent purity and yield obtained in one step. This resin comes with complete technical and regulatory support.
Main characteristics
- Matrix: cross-linked poly(styrene-divinylbenzene) POROS beads
- Average particle size: 50 μm
- Ligand: CaptureSelect AAVX affinity ligand
- Binding capacity: >1 x 1013 genome copies/mL of resin
- Elution conditions: acidic elution with 0.1 M glycine, pH 3.0
- Mechanical resistance: 100 bar (1450 psi; 10 MPa)
- Formulation buffer: 18% (v/v) ethanol
Free of animal components
CaptureSelect products contain affinity ligands based on recombinant single-domain antibody fragments (VHH) created by a proprietary technology. The VHH affinity ligand is a 12 to 15 kDa fragment comprising the three complementarity-determining regions (CDRs) that form the antigen-binding domain, efficiently produced in the yeast Saccharomyces cerevisiae by a production process free of any animal components (Animal Origin-Free).
Specifications
>1 x 1013 genome copies/mL of resin | |
Wet Ice | |
Liquid Suspension | |
Resin |
Store in refrigerator at 2°C to 8°C. | |
50 μm | |
POROS™ | |
Affinity |