Learn More
Extech Portable Indoor Air Quality CO2 Meter
Use the Portable Indoor Air Quality CO2 Meter with built-in RS-232 interface for capturing readings on your PC.
Brand:  Extech 81973-90
Product Code. 12200419
  • £695.00 / Each
Explore more special offers
Description
Description
  • Check air quality in schools, office buildings, greenhouses, and anywhere that high levels of carbon dioxide are generated.
  • Features a maintenance-free NDIR (nondispersive infrared) CO2 sensor, user-programmable audible alarm, backlit LCD, and RS-232 interface.
  • Meter calculates statistical weighted averages of TWA (8 hour time-weighted average) and STEL (15 minutes short term exposure limit).
Specifications
Specifications
CO2
14°F to 140°F
Audible
Backlit LED
RS-232
Portable Indoor Air Quality CO2 Meter
-10°C to +60°C
RS-232
0 to 5000 ppm (CO2), 0.0 to 99.9% (RH)
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Extech Portable Indoor Air Quality CO2 Meter > Portable Indoor Air Quality CO2 Meter; Measurement Range: 0 to 5000 ppm (CO2), 0.0 to 99.9% (RH)