Learn More
TFA Dostmann CO2 Monitor AIRCO2NTROL COACH
The CO2 Monitor AIRCO2NTROL COACH shows the current CO2 content of the room air, the temperature, humidity, and the time. Depending on the settings, it displays the current CO2 readings through the traffic light colours. The graph clearly shows the CO2 levels development of the last 24 hours.
Brand:  TFA Dostmann 31.5009.02
Product Code. 17941662
  • £92.35 / Each
Explore more special offers
Description
Description
Who breathes good air is healthier, happier and can do more. Headaches, dizziness and permanent fatigue are often due to a high concentration of CO2 in the air. TFA's AIRCO2NTROL COACH CO2 monitor helps you monitor how stale and contaminated the air is in indoor spaces where many people are present such as in schools, offices and public buildings. Timely and energy-efficient ventilation ensures a healthy indoor climate. Furthermore, a high CO2 value is considered an indicator for a possibly virus-contaminated aerosol concentration in the room air. The more exhaled air there is in the room, the higher the probability of infection via aerosols. The monitor shows on the display the current CO2 content of the room air, the temperature, humidity and the time.
The display's illumination color can be used to quickly evaluate and interpret the current CO2 readings. Depending on the setting, the illumination directly signals the current air quality in the traffic light colors (green, orange, red). Alternatively, the red display illumination can be used as an optical signal in the event of an alarm when a certain limit has been exceeded. As a further variant, you can choose a solid color from seven shades or switch off the illumination completely.
The three smiley icons serve as a comfort-level zone indicator for the measured values of the last 24 hours. If the reading is at the level of the happy smiley, the air quality is ideal to good. In addition, the graph clearly shows the CO2 levels development of the last 24 hours. The power is supplied via a suitable USB power source (PC, laptop).
  • High-quality NDIR Dual Beam Sensor
  • CO2 level measurement with 24-hours graph
  • Micro-USB cable inclusive
  • For the monitoring of CO2 concentration in buildings, where people are present, e.g. schools, offices, public buildings
  • Backlight color shows CO2 level, alarm or preferred color
  • Indication of time
  • Indoor temperature and humidity
Specifications
Specifications
Carbon Dioxide
5 V
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
TFA Dostmann CO2 Monitor AIRCO2NTROL COACH > Detectable Analytes: Carbon Dioxide