Learn More
PSF Progressive Safety™ Portwest™ Antistatic Laced Janitorial Boot
Providing water resistant safety at an affordable price.
Brand:  PSF Progressive Safety™ FW88-11
Code : NEW
Additional Details : Weight : 0.50000kg
 View more versions of this product
Product Code. 15669578
  • £41.70 / 1 pair
Estimated Shipment: 08-08-2024
to see stock.
Add to basket
Description
Description
  • All safety footwear is designed with both comfort and durability in mind
  • Conforms to EN ISO 20345:2011 for 200 joules protection
  • Water resistant upper
  • Antistatic and antislip dual density outsole
  • Laced version ideal for janitorial work
  • Antibacterial lining
  • CE certified
Specifications
Specifications
EN ISO 20345:2011
Laced boot
Microfiber
Energy absorbing, slip-resistant
White
Janitorial work
11
Water, oil, slip-resistant