missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ Positive Displacement Pipette Tips
Contains replacement consumables to run all Saftest™ test kits.
Brand: MP Biomedicals™ 07DP1038
106.91 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- For use with SafTest analyzer
- Used with all six SafTest test kits
- Appropriate for all testing endpoints
- Research area: Lipid degradation and oxidation
- Detection method: Bichromatic photometer
- Application area: Oil/fat degradation
- Construction: Polystryene
- Pre assembled, pre sterilized

Specifications
apillaries and pistons are pre-assembled and presterilized | |
Sterile | |
Polypropylene |
200 to 1000 μL | |
SafTest Analyzer |
Special Offers
-
Multiple Manufacturers
Contract Testing Lab
Use promotion code 24138 to get amazing savings on products for your analysisPromo Code: 24138Expires: 2025-04-30
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
MP Biomedicals™ Positive Displacement Pipette Tips > Capacity: 200 to 1000μL
Spot an opportunity for improvement?Share a Content Correction