missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium acetate, 1M aq. soln., pH 7.5, Thermo Scientific™
£34.60 - £53.60
Chemical Identifiers
CAS | 127-08-2 |
---|---|
Molecular Formula | C2H3KO2 |
Molecular Weight (g/mol) | 98.142 |
MDL Number | MFCD00012458 |
InChI Key | SCVFZCLFOSHCOH-UHFFFAOYSA-M |
Synonym | potassium acetate, acetic acid, potassium salt, diuretic salt, potassium ethanoate, koac, octan draselny czech, potassiumacetate, acetic acid potassium salt, acok, fema no. 2920 |
PubChem CID | 517044 |
ChEBI | CHEBI:32029 |
IUPAC Name | potassium;acetate |
SMILES | CC(=O)[O-].[K+] |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
127-08-2 | |
98.142 | |
SCVFZCLFOSHCOH-UHFFFAOYSA-M | |
517044 | |
potassium;acetate |
C2H3KO2 | |
MFCD00012458 | |
potassium acetate, acetic acid, potassium salt, diuretic salt, potassium ethanoate, koac, octan draselny czech, potassiumacetate, acetic acid potassium salt, acok, fema no. 2920 | |
CHEBI:32029 | |
CC(=O)[O-].[K+] |
Specifications
Colorless | |
C2H3KO2 | |
MFCD00012458 | |
potassium acetate, acetic acid, potassium salt, diuretic salt, potassium ethanoate, koac, octan draselny czech, potassiumacetate, acetic acid potassium salt, acok, fema no. 2920 | |
CC(=O)[O-].[K+] | |
98.142 | |
CHEBI:32029 | |
7.5 | |
Potassium acetate |
127-08-2 | |
CH3CO2K | |
1M aq. soln. | |
SCVFZCLFOSHCOH-UHFFFAOYSA-M | |
potassium;acetate | |
517044 | |
98.15 | |
Liquid |
Safety and Handling
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures