missing translation for 'onlineSavingsMsg'
Learn More

Potassium antimonyl tartrate sesquihydrate, 99+%, Thermo Scientific Chemicals

£24.72 - £324.62

Chemical Identifiers

CAS 28300-74-5
Molecular Formula 1·5 H2O
Molecular Weight (g/mol) 333.93
MDL Number MFCD00167056
Synonym Antimonyl potassium tartrate sesquihydrate
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10174800
View Documents Promotion Details
Thermo Scientific Acros
223801000
100 g Glass bottle
£24.72
100g
Please to purchase this item. Need a web account? Register with us today!
10772841
View Documents Promotion Details
Thermo Scientific Acros
223805000
500 g Glass bottle
£91.25
500g
Please to purchase this item. Need a web account? Register with us today!
10105363
View Documents Promotion Details
Thermo Scientific Acros
223800025
2.5 kg Glass bottle
£324.62
2.50kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

28300-74-5
333.93
Antimonyl potassium tartrate sesquihydrate
1·5 H2O
MFCD00167056

Specifications

100.0°C
White
99% min. (Iodometry)
MFCD00167056
Antimonyl potassium tartrate sesquihydrate
333.93
≥99%
2.7% max. (105°C)
28300-74-5
Fine Crystalline Powder
1·5 H2O
15, 691
Solubility in water: 8.3%. Other solubilities: 1g/3mL boiling water, 1g/15mL glycerol, insoluble in alcohol
333.93
Authentic
Potassium antimonyl tartrate sesquihydrate, 99+%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Toxic to aquatic life with long lasting effects.
Harmful if inhaled.
Harmful if swallowed.

GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Avoid release to the environment.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

GHS Signal Word: Warning

missing translation for 'rtecsNumber' : CC6825000

Product Identifier
  • Potassium antimonyl tartrate sesquihydrate

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 2

Hazard Statement
  • H302-Harmful if swallowed.
  • H332-Harmful if inhaled.
  • H411-Toxic to aquatic life with long lasting effects.

Precautionary Statement
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P264-Wash thoroughly after handling.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only