missing translation for 'onlineSavingsMsg'
Learn More

Potassium Bicarbonate, ≥99.95% Trace Metals Basis, Honeywell™

≥99.95% trace metals basis, 99.7-100.5% dry basis

£89.92 - £340.87

Chemical Identifiers

CAS 298-14-6
Molecular Formula CHKO3
Molecular Weight (g/mol) 100.11
MDL Number MFCD00011402
InChI Key TYJJADVDDVDEDZ-UHFFFAOYSA-M
Synonym potassium bicarbonate, potassium hydrogen carbonate, potassium hydrogencarbonate, potassium acid carbonate, monopotassium carbonate, carbonic acid, monopotassium salt, unii-hm5z15lebn, potassiumbicarbonate, ccris 3510, khco3
PubChem CID 516893
ChEBI CHEBI:81862
IUPAC Name potassium;hydrogen carbonate
SMILES [K+].OC([O-])=O
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
15675510
View Documents
Honeywell Research Chemicals
431583-50G
50 g Poly Bottle
£89.92
50g
Please to purchase this item. Need a web account? Register with us today!
15665510
View Documents
Honeywell Research Chemicals
431583-250G
250 g Poly Bottle
£340.87
250g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

  • We are your supplier of high-quality lab reagents and chemicals for analytical and chemistry laboratories:
    Solvents – ACS specific use (HPLC), ACS general use, solvents for histology, and reagent grade for chemical synthesis and other industrial applications.
    Inorganics – Chemical synthesis and inorganic chemistry including essential acids and bases, salts, metals and elements, and reagents for chemical reactions.
Specifications

Chemical Identifiers

298-14-6
100.11
TYJJADVDDVDEDZ-UHFFFAOYSA-M
516893
potassium;hydrogen carbonate
CHKO3
MFCD00011402
potassium bicarbonate, potassium hydrogen carbonate, potassium hydrogencarbonate, potassium acid carbonate, monopotassium carbonate, carbonic acid, monopotassium salt, unii-hm5z15lebn, potassiumbicarbonate, ccris 3510, khco3
CHEBI:81862
[K+].OC([O-])=O

Specifications

100°C to 120°C
White
CHKO3
MFCD00011402
4535309
In water 333g/L at 20°C
[K+].OC([O-])=O
100.11
CHEBI:81862
≥99.95% (trace metals basis)
Powder
Potassium bicarbonate
298-14-6
8 to 8.6 (at 20°C)
KHCO3
NONH for all modes of transport
potassium bicarbonate, potassium hydrogen carbonate, potassium hydrogencarbonate, potassium acid carbonate, monopotassium carbonate, carbonic acid, monopotassium salt, unii-hm5z15lebn, potassiumbicarbonate, ccris 3510, khco3
TYJJADVDDVDEDZ-UHFFFAOYSA-M
potassium;hydrogen carbonate
516893
100.12g/mol
Odorless
2.17g/cm3 at 20°C
Videos
Safety and Handling

Safety and Handling

missing translation for 'rtecsNumber' : 206-059-0

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.