Learn More
Potassium bis(oxalato)oxotitanate(IV) dihydrate, Thermo Scientific Chemicals
Mordant in dyeing, reagent
£60.30 - £208.00
Chemical Identifiers
CAS | 14402-67-6 |
---|---|
Molecular Formula | C4K2O9Ti |
Molecular Weight (g/mol) | 318.10 |
MDL Number | MFCD00150592 |
InChI Key | UHWHMHPXHWHWPX-UHFFFAOYSA-J |
Synonym | Titanium potassium oxalate |
IUPAC Name | dipotassium oxotitaniumbis(ylium) dioxalate |
SMILES | [K+].[K+].O=[Ti++].[O-]C(=O)C([O-])=O.[O-]C(=O)C([O-])=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11443800
|
Thermo Scientific Alfa Aesar
|
500 g |
£60.30
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11433800
|
Thermo Scientific Alfa Aesar
|
2 kg |
£208.00
2kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Mordant in dyeing, reagent Potassium bis(oxalato)oxotitanate(IV) dihydrate is used as a mordant in dyeing and acts as an analytical reagent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
14402-67-6 | |
318.10 | |
UHWHMHPXHWHWPX-UHFFFAOYSA-J | |
dipotassium oxotitaniumbis(ylium) dioxalate |
C4K2O9Ti | |
MFCD00150592 | |
Titanium potassium oxalate | |
[K+].[K+].O=[Ti++].[O-]C(=O)C([O-])=O.[O-]C(=O)C([O-])=O |
Specifications
14402-67-6 | |
Odorless | |
MFCD00150592 | |
Titanium potassium oxalate | |
UHWHMHPXHWHWPX-UHFFFAOYSA-J | |
dipotassium oxotitaniumbis(ylium) dioxalate | |
354.10 (318.10 Anhydrous) | |
Potassium bis (oxalato)oxotitanate (IV) dihydrate |
Colorless | |
C4K2O9Ti | |
UN3288 | |
Soluble in water. | |
[K+].[K+].O=[Ti++].[O-]C(=O)C([O-])=O.[O-]C(=O)C([O-])=O | |
318.10 | |
Crystalline |
Safety and Handling
GHS H Statement
H302-H312
Harmful if swallowed.
Harmful in contact with skin.
P264b-P270-P280-P301+P312-P302+P352-P312-P330-P363-P501c
H302+H312
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLID, INORGANIC, N.O.S.
missing translation for 'einecsNumber' : 238-475-3
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only