missing translation for 'onlineSavingsMsg'
Learn More
Potassium borohydride, 98+%, Thermo Scientific Chemicals Product Code.: 10123880

Potassium borohydride, 98+%, Thermo Scientific Chemicals

Product Code. 10123880
500 g, Glass bottle
Click to view available options
Quantity:
5 g
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Plastic drum
This item is not returnable. View return policy

Product Code. 10123880

Brand: Thermo Scientific Acros 196555000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Reduction reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13762-51-1
Molecular Formula BH4K
Molecular Weight (g/mol) 53.94
MDL Number MFCD00011396
InChI Key ICRGAIPBTSPUEX-UHFFFAOYSA-N
Synonym potassium borohydride, potassium tetrahydroborate, potassium borohydrate, unii-0ws230dtgf, potassium tetrahydroborate 1-, borohydrure de potassium french, borate 1-, tetrahydro-, potassium, 0ws230dtgf, borohydrure de potassium, borylpotassiumShow MoreShow Less
PubChem CID 22892188
SMILES [BH4-].[K+]

Specifications

Melting Point 500.0°C
Color White
Physical Form Crystalline Powder
Quantity 500 g
Assay Percent Range 98+%
Linear Formula KBH4
Fieser 04,398; 06,476; 08,407
Merck Index 15, 7737
Solubility Information Solubility in water: 190g/L (25°C). Other solubilities: reacts with water, partially soluble in ammonia, insoluble in ethers, hydrocarbons, isopropylamine,, benzene, hexane, dioxane, thf and acetonitrile, 7g/L methanol (20°C)
IUPAC Name potassium;boron(1-)
Formula Weight 53.93
Percent Purity ≥98%
Packaging Glass bottle
Chemical Name or Material Potassium borohydride, 98%
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0471841 Certificate TypeCertificate of Analysis Date10/04/2025 Catalog Number196551000,196550050,196555000,196550025
Lot NumberA0466493 Certificate TypeCertificate of Analysis Date11/10/2024 Catalog Number196551000,196550050,196555000,196550025
Lot NumberA0464161 Certificate TypeCertificate of Analysis Date28/06/2024 Catalog Number196551000,196550050,196555000,196550025
Lot NumberA0458008 Certificate TypeCertificate of Analysis Date23/11/2023 Catalog Number196551000,196550050,196555000,196550025
Lot NumberA0410286 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number196551000,196550050,196555000,196550025

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Potassium borohydride, 98+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.