missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium borohydride, 98+%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Plastic drum
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reduction reagent

Specifications
Specifications
Melting Point | 500.0°C |
Color | White |
Physical Form | Crystalline Powder |
Quantity | 100 g |
Assay Percent Range | 98+% |
Linear Formula | KBH4 |
Fieser | 04,398; 06,476; 08,407 |
Merck Index | 15, 7737 |
Solubility Information | Solubility in water: 190g/L (25°C). Other solubilities: reacts with water, partially soluble in ammonia, insoluble in ethers, hydrocarbons, isopropylamine,, benzene, hexane, dioxane, thf and acetonitrile, 7g/L methanol (20°C) |
IUPAC Name | potassium;boron(1-) |
Show More |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
A0466493 | Certificate of Analysis | 11/10/2024 | 196551000,196550050,196555000,196550025 |
A0464161 | Certificate of Analysis | 28/06/2024 | 196551000,196550050,196555000,196550025 |
A0458008 | Certificate of Analysis | 23/11/2023 | 196551000,196550050,196555000,196550025 |
A0410286 | Certificate of Analysis | 25/08/2023 | 196551000,196550050,196555000,196550025 |
A0455563 | Certificate of Analysis | 25/08/2023 | 196551000,196550050,196555000,196550025 |
Safety and Handling



Product Identifier
- Potassium borohydride
- Danger
- Acute toxicity Category 3
- Skin corrosion/irritation Category 1 B
- Substance which\, in contact with water\, emits flammable gases Category 2
- H261-In contact with water releases flammable gases.
- H301-Toxic if swallowed.
- H311-Toxic in contact with skin.
- H314-Causes severe skin burns and eye damage.
- EUH014-Reacts violently with water.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P402+P404-Store in a dry place. Store in a closed container.
- MIXTURE LIST-Contains : Potassium borohydride
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Potassium borohydride, 98+%, Thermo Scientific Chemicals >
Spot an opportunity for improvement?Share a Content Correction