missing translation for 'onlineSavingsMsg'
Learn More

Potassium chlorate, 99+%, ACS reagent, Thermo Scientific Chemicals

£25.53 - £189.95

Chemical Identifiers

CAS 3811-04-9
Molecular Formula ClKO3
Molecular Weight (g/mol) 122.55
MDL Number MFCD00011361
InChI Key VKJKEPKFPUWCAS-UHFFFAOYSA-M
Synonym potassium chlorate, fekabit, potcrate, kaliumchlorat, anforstan, berthollet salt, salt of tarter, berthollet's salt, chlorate of potash, chloric acid, potassium salt
PubChem CID 6426889
IUPAC Name potassium;chlorate
SMILES [O-]Cl(=O)=O.[K+]
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10076010
View Documents
Thermo Scientific Acros
418190050
5 g Glass Bottle
£25.53
5g
Please to purchase this item. Need a web account? Register with us today!
10259370
View Documents
Thermo Scientific Acros
418195000
500 g Plastic bottle
£54.27
500g
Please to purchase this item. Need a web account? Register with us today!
10227563
View Documents
Thermo Scientific Acros
418190025
2.5 kg Plastic bottle
£189.95
2.50kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Oxidation reagent
Specifications

Chemical Identifiers

3811-04-9
122.55
VKJKEPKFPUWCAS-UHFFFAOYSA-M
6426889
[O-]Cl(=O)=O.[K+]
ClKO3
MFCD00011361
potassium chlorate, fekabit, potcrate, kaliumchlorat, anforstan, berthollet salt, salt of tarter, berthollet's salt, chlorate of potash, chloric acid, potassium salt
potassium;chlorate

Specifications

356.0°C to 368.0°C
White
99+%
KClO3
01,927
potassium chlorate, fekabit, potcrate, kaliumchlorat, anforstan, berthollet salt, salt of tarter, berthollet's salt, chlorate of potash, chloric acid, potassium salt
VKJKEPKFPUWCAS-UHFFFAOYSA-M
potassium;chlorate
6426889
≥99%
Potassium chlorate
3811-04-9
Crystalline Powder
ClKO3
MFCD00011361
15, 7741
Solubility in water: 73g/L (20°C). Other solubilities: 555g/L hot water (100°C), soluble in alkalies, 1g/50mL glycerol, practically insoluble in alcohol, insoluble in ether
[O-]Cl(=O)=O.[K+]
122.55
122.55
ACS Reagent
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Toxic to aquatic life with long lasting effects.
Harmful if swallowed.
Harmful if inhaled.
May cause fire or explosion; strong oxidizer.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Take any precaution to avoid mixing with combustibles.
In case of major fire and large quantities: Evacuate area.
Fight fire remotely due to the ris

GHS Signal Word: Danger

missing translation for 'einecsNumber' : 223-289-7

missing translation for 'rtecsNumber' : FO0350000

missing translation for 'tsca' : TSCA

Product Identifier
  • Potassium chlorate

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Oxidising solid Category 1

Hazard Statement
  • H411-Toxic to aquatic life with long lasting effects.
  • H302-Harmful if swallowed.
  • H332-Harmful if inhaled.
  • H271-May cause fire or explosion; strong oxidiser.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P371+P380+P375-In case of major fire and large quantities: Evacuate area Fight fire remotely due to the risk of explosion.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only