missing translation for 'onlineSavingsMsg'
Learn More
Potassium chloride, 99+%, for analysis, Thermo Scientific Chemicals Product Code.: 10236350

Potassium chloride, 99+%, for analysis, Thermo Scientific Chemicals

Product Code. 10236350
1 kg, Plastic bottle
Click to view available options
Quantity:
1 kg
5 kg
25 kg
Packaging:
Fibre drum
Plastic bottle
This item is not returnable. View return policy

Product Code. 10236350

Brand: Thermo Scientific Acros 196770010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Biological buffer for analysis
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7447-40-7
Molecular Formula ClK
Molecular Weight (g/mol) 74.55
MDL Number MFCD00011360
InChI Key WCUXLLCKKVVCTQ-UHFFFAOYSA-M
Synonym potassium chloride, enseal, muriate of potash, klotrix, sylvite, slow-k, potassium chloride kcl, klor-con, chlorvescent, kalitabs
PubChem CID 4873
ChEBI CHEBI:32588
SMILES [Cl-].[K+]

Specifications

Boiling Point 1420.0°C
Melting Point 770.0°C
Color Colorless
Physical Form Liquid
Quantity 1 kg
Assay Percent Range 99.0 to 100.5% (on dried substance) (Argentometry)
Linear Formula KCl
Fieser 06,48
Merck Index 15,7742
Solubility Information Solubility in water: 340g/L (20°C). Other solubilities: slightly soluble in alcohol, insoluble in ether and acetone
IUPAC Name potassium chloride
Formula Weight 74.54
Percent Purity ≥99%
Grade Analytical
Loss on Drying 0.5 % max. (1 g, 105°C, 3 hrs)
Packaging Plastic bottle
Chemical Name or Material Potassium chloride, p.a.
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0467308 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number196770010,196770050,196770250
Lot NumberA0466102 Certificate TypeCertificate of Analysis Date18/09/2024 Catalog Number196770010,196770050,196770250
Lot NumberA0463113 Certificate TypeCertificate of Analysis Date17/06/2024 Catalog Number196770010,196770050,196770250
Lot NumberA0462228 Certificate TypeCertificate of Analysis Date25/04/2024 Catalog Number196770010,196770050,196770250
Lot NumberA0452447 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number196770010,196770050,196770250

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Potassium chloride, 99+%, for analysis, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.