Learn More

- Home
- Products
- Chemicals
- Salts and Inorganics
- Inorganic Salts
- Alkali Metal Salts
- Potassium chloride, 99+%, ACS reagent, Thermo Scientific Chemicals
Potassium chloride, 99+%, ACS reagent, Thermo Scientific Chemicals
Product Code. 10730492Product Code. 10730492
Brand: Thermo Scientific Acros 424090010
Use promo code "24155" to get your promotional price.
Explore more special offers
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Chemical Identifiers
CAS | 7447-40-7 |
Molecular Formula | ClK |
Molecular Weight (g/mol) | 74.55 |
MDL Number | MFCD00011360 |
InChI Key | WCUXLLCKKVVCTQ-UHFFFAOYSA-M |
Synonym | potassium chloride, enseal, muriate of potash, klotrix, sylvite, slow-k, potassium chloride kcl, klor-con, chlorvescent, kalitabs |
PubChem CID | 4873 |
ChEBI | CHEBI:32588 |
SMILES | [Cl-].[K+] |
Specifications
Color | White |
pH | 5.4 to 8.6 (5% soln. at 25°C) |
Physical Form | Crystalline Powder and/or Crystals |
Quantity | 1 kg |
Assay Percent Range | 99+% |
Linear Formula | KCl |
Fieser | 06,480 |
Merck Index | 15, 7742 |
IUPAC Name | potassium;chloride |
Formula Weight | 74.54 |
Percent Purity | 99.0 to 100.5% |
Grade | ACS Reagent |
Packaging | Plastic bottle |
Chemical Name or Material | Potassium Chloride |
Show More Show Less |
Product Suggestions
You may be interested in these alternatives
-
Savings of £0.78Product Code. 15675920
-
-
Product Code. 11347337
-
Product Code. 10236350
-
Product Code. 15691160
-
-
-
-
Product Code. 10208673
Customers who viewed this item also viewed. This information does not imply a recommendation or representation of any kind and any action taken upon the information provided is strictly at your own risk. The products shown have not been qualified by Fisher Scientific for any particular purpose, application or use and it is the responsibility of the purchaser to ensure that the performance of the product is suitable for the purchasers specific use or intended application.
-
Catalog No. 11444783
£29.42 / 1000g -
Catalog No. 10236350
£60.17 / 1kg -
Catalog No. 10375810
£265.00 / 500g -
Catalog No. 10404385
£35.33 / 1kg -
Catalog No. 10735874
£48.95 / 500g -
Catalog No. 10112640
£50.40 / 1kg -
Catalog No. 10703811
£258.00 / 500g -
Catalog No. 10793611
£101.00 / 1kg -
Catalog No. 10117113
£25.71 / 100g
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
Lot NumberA0467626 | Certificate TypeCertificate of Analysis | Date31/01/2025 | Catalog Number424090010,424090250,424090050 |
Lot NumberB0156108 | Certificate TypeCertificate of Analysis | Date19/12/2024 | Catalog Number424090010,424090250,424090050 |
Lot NumberA0461703 | Certificate TypeCertificate of Analysis | Date12/04/2024 | Catalog Number424090010,424090250,424090050 |
Lot NumberA0461018 | Certificate TypeCertificate of Analysis | Date14/03/2024 | Catalog Number424090010,424090250,424090050 |
Lot NumberB0154439 | Certificate TypeCertificate of Analysis | Date05/12/2023 | Catalog Number424090010,424090250,424090050 |
- 1
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.