missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Chloride, Crystal, U.S.P. - F.C.C., J.T. Baker™
£118.00 - £303.00
Chemical Identifiers
CAS | 7447-40-7 |
---|---|
Molecular Formula | ClK |
Molecular Weight (g/mol) | 74.55 |
MDL Number | MFCD00011360 |
InChI Key | WCUXLLCKKVVCTQ-UHFFFAOYSA-M |
Synonym | potassium chloride, enseal, muriate of potash, klotrix, sylvite, slow-k, potassium chloride kcl, klor-con, chlorvescent, kalitabs |
PubChem CID | 4873 |
ChEBI | CHEBI:32588 |
SMILES | [Cl-].[K+] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
15578074
|
Avantor J.T.Baker
3046-01 |
500 g | Poly Bottle |
£118.00
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15588074
|
Avantor J.T.Baker
3046-05 |
2.5 kg | Poly Bottle |
£303.00
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Chemical Identifiers
7447-40-7 | |
74.55 | |
WCUXLLCKKVVCTQ-UHFFFAOYSA-M | |
4873 | |
[Cl-].[K+] |
ClK | |
MFCD00011360 | |
potassium chloride, enseal, muriate of potash, klotrix, sylvite, slow-k, potassium chloride kcl, klor-con, chlorvescent, kalitabs | |
CHEBI:32588 |
Specifications
7447-40-7 | |
ClK | |
potassium chloride, enseal, muriate of potash, klotrix, sylvite, slow-k, potassium chloride kcl, klor-con, chlorvescent, kalitabs | |
[Cl-].[K+] | |
74.55 | |
CHEBI:32588 | |
FCC/USP | |
Potassium Chloride |
Crystals | |
MFCD00011360 | |
WCUXLLCKKVVCTQ-UHFFFAOYSA-M | |
potassium;chloride | |
4873 | |
74.55 | |
1L = 1.99kg |