missing translation for 'onlineSavingsMsg'
Learn More

Potassium hexahydroxoantimonate(V), Honeywell Fluka™

for the precipitation of sodium,≥99.0%

£60.79 - £243.97

Chemical Identifiers

CAS 12208-13-8
MDL Number MFCD00011415
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15605990
View Documents Promotion Details
Honeywell-Fluka
60500-100G
100 g
£60.79
100g
Please to purchase this item. Need a web account? Register with us today!
15615990
View Documents Promotion Details
Honeywell-Fluka
60500-500G
500 g
£243.97
500g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications

Chemical Identifiers

12208-13-8
MFCD00011415

Specifications

12208-13-8
MFCD00011415
262.9g/mol
Potassium hexahydroxoantimonate(V)
KSb(OH)6
UN1549
≥99%
Videos
Safety and Handling

Safety and Handling

P273

Product Identifier
  • Potassium hexahydroxoantimonate(V)

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 2

Hazard Statement
  • H302-Harmful if swallowed.
  • H332-Harmful if inhaled.
  • H411-Toxic to aquatic life with long lasting effects.

Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

Supplemental information
  • MIXTURE LIST-Contains : potassium hexahydroxoantimonate

SDS
Documents

Documents

Product Certifications