missing translation for 'onlineSavingsMsg'
Learn More

Potassium hydrogen carbonate, ACS reagent, Thermo Scientific Chemicals

£17.69 - £160.80

Chemical Identifiers

CAS 298-14-6
Molecular Formula CHKO3
Molecular Weight (g/mol) 100.11
MDL Number MFCD00011402
InChI Key TYJJADVDDVDEDZ-UHFFFAOYSA-M
Synonym potassium bicarbonate, potassium hydrogen carbonate, potassium hydrogencarbonate, potassium acid carbonate, monopotassium carbonate, carbonic acid, monopotassium salt, unii-hm5z15lebn, potassiumbicarbonate, ccris 3510, khco3
PubChem CID 516893
ChEBI CHEBI:81862
SMILES [K+].OC([O-])=O
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11914311
View Documents Promotion Details
Thermo Scientific Acros
446301000
100 g
£17.69
100g
Please to purchase this item. Need a web account? Register with us today!
11934311
View Documents Promotion Details
Thermo Scientific Acros
446305000
500 g
£53.97
500g
Please to purchase this item. Need a web account? Register with us today!
11924311
View Documents
Thermo Scientific Acros
446300025
2.5 kg
£160.80
2.50kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

298-14-6
100.11
TYJJADVDDVDEDZ-UHFFFAOYSA-M
516893
[K+].OC([O-])=O
CHKO3
MFCD00011402
potassium bicarbonate, potassium hydrogen carbonate, potassium hydrogencarbonate, potassium acid carbonate, monopotassium carbonate, carbonic acid, monopotassium salt, unii-hm5z15lebn, potassiumbicarbonate, ccris 3510, khco3
CHEBI:81862

Specifications

292.0°C
CHKO3
MFCD00011402
potassium bicarbonate, potassium hydrogen carbonate, potassium hydrogencarbonate, potassium acid carbonate, monopotassium carbonate, carbonic acid, monopotassium salt, unii-hm5z15lebn, potassiumbicarbonate, ccris 3510, khco3
TYJJADVDDVDEDZ-UHFFFAOYSA-M
potassium;hydrogen carbonate
516893
100.12
Plastic bottle
298-14-6
KHCO3
15,773
Solubility in water: 333g/L (20°C). Other solubilities: 600g/L water (60°C), practocally insoluble in alcohol, soluble in potassium carbonate solution
[K+].OC([O-])=O
100.11
CHEBI:81862
ACS Reagent
Potassium hydrogen carbonate
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 206-059-

Product Identifier
  • Potassium hydrogen carbonate

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only