Learn More
Potassium hydrogen fluoride, 99+%, extra pure, Thermo Scientific Chemicals
£21.00 - £133.00
Chemical Identifiers
CAS | 7789-29-9 |
---|---|
Molecular Formula | HF2K |
Molecular Weight (g/mol) | 78.1 |
MDL Number | MFCD00011403 |
Synonym | Potassium bifluoride |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10761491
|
Thermo Scientific Acros
219230250 |
25 g | Plastic bottle |
£21.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10134990
|
Thermo Scientific Acros
219235000 |
500 g | Plastic bottle |
£40.50
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10236610
|
Thermo Scientific Acros
219230025 |
2.5 kg | Plastic bottle |
£133.00
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagentChemical Identifiers
7789-29-9 | |
78.1 | |
Potassium bifluoride |
HF2K | |
MFCD00011403 |
Specifications
239.0°C | |
White to Gray | |
99+% | |
KHF2 | |
1811 | |
15, 7731 | |
Solubility in water: 39g/100mL (20°C). Other solubilities: soluble in diluted acids, insoluble in absolute alcohol, soluble in diluted alcohol | |
78.1 | |
Extra Pure |
7789-29-9 | |
Crystalline Powder and/or Crystals | |
HF2K | |
MFCD00011403 | |
16,283 | |
Potassium bifluoride | |
78.1 | |
≥99% | |
Potassium hydrogen fluoride, 99+% |
Safety and Handling
GHS H Statement:
Causes severe skin burns and eye damage.
Toxic if swallowed.
GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Wear eye protection/face protection.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 232-156-2
missing translation for 'rtecsNumber' : TS6650000
missing translation for 'tsca' : TSCA
- Potassium hydrogen fluoride
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- Skin corrosion/irritation Category 1 B
- Serious eye damage/eye irritation Category 1
Hazard Statement
- H314-Causes severe skin burns and eye damage.
- H301-Toxic if swallowed.
Precautionary Statement
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
RUO – Research Use Only