Learn More
Potassium hydroxide, ACS, 85% min, K{2}CO{3} 2.0% max, Thermo Scientific Chemicals
Description
In soap manufacture, in bleaching, as an electrolyte, as an absorbent, in dyestuffs, in liquid fertilizer, in electroplating, as a reagentPotassium hydroxide acts as a precursor for most of the soft and liquid soaps, and for numerous potassium-containing chemicals. It is widely used in the manufacture of detergents, fertilizer, rayon, paper, dyestuffs, various drugs, drug intermediates, fine chemicals, and petroleum products. In the field of organic chemistry, potassium hydroxide acts as a base, and is used in the synthesis of several organic compounds. Further, it finds application as an electrolyte in alkaline batteries, which are based on nickel-cadmium, nickel-hydrogen and manganese dioxide-zinc.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Boiling Point | 1320°C to 1324°C |
Melting Point | 360°C |
Color | White |
Physical Form | Pellets |
Quantity | 10 kg |
Assay Percent Range | 85% min. |
Solubility Information | Soluble in water,alcohol,and glycerol. Slightly soluble in ether. |
IUPAC Name | potassium hydroxide |
Formula Weight | 56.11 |
Grade | ACS |
Show More |
Product Suggestions
Customers who viewed this item also viewed.
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
N04L011 | Certificate of Analysis | 06/02/2025 | 013451.A9,013451.A3,013451.36,013451.18 |
M13L010 | Certificate of Analysis | 15/01/2025 | 013451.A9,013451.A3,013451.36,013451.18 |
Z10K030 | Certificate of Analysis | 11/12/2024 | 013451.A9,013451.A3,013451.36,013451.18 |
W03K002 | Certificate of Analysis | 11/09/2024 | 013451.A9,013451.A3,013451.36,013451.18 |
U27K034 | Certificate of Analysis | 11/09/2024 | 013451.A9,013451.A3,013451.36,013451.18 |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.