missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Hydroxide, Certified AR for Analysis, Pellets, meets analytical specification of Ph.Eur., BP, Fisher Chemical™
Click to view available options
Quantity:
500 g
1 kg
5 kg
Description
HKO, CAS Number-1310-58-3, potash lye, caustic potash, hydroxyde de potassium, caswell no. 693, potassium hydrate, potassium hydroxide, potasse caustique, potassa, potassium hydroxide solution, potassium hydroxide k oh, 500g, 1320 deg.C, CHEBI:32035, White, 56.11g/mol, KWYUFKZDYYNOTN-UHFFFAOYSA-M

Specifications
Specifications
pH | 13.5 |
Color | White |
Boiling Point | 1320°C |
Melting Point | 360°C |
Quantity | 5 kg |
IUPAC Name | potassium hydroxide |
Formula Weight | 56.11g/mol |
Packaging | Plastic powder jar |
Physical Form | Solid |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Safety and Handling


Product Identifier
- Potassium hydroxide
- Danger
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 1 A
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H302-Harmful if swallowed.
- H290-May be corrosive to metals.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Potassium Hydroxide, Certified AR for Analysis, Pellets, meets analytical specification of Ph.Eur., BP, Fisher Chemical™ >
Spot an opportunity for improvement?Share a Content Correction