missing translation for 'onlineSavingsMsg'
Learn More
Sodium hydroxide, pellets, 98% Product Code.: 15433117

Sodium hydroxide, pellets, 98%

Product Code. 15433117
500 g
Click to view available options
Quantity:
500 g
This item is not returnable. View return policy

Product Code. 15433117

Brand: Thermo Scientific Alfa Aesar A16037.36

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1310-73-2

Sodium hydroxide is widely used in variety of large scale applications such as pulp and paper manufacturing, in the production of soap and detergents as well as water treatment. It finds application in the base- driven hydrolysis of esters, amides and alkyl halides. It is also used as an industrial cleaning agent, an additive in drilling mud to increase alkalinity in bentonite in petroleum industry and as a powerful degreaser on stainless steel and glass bakeware. It is utilized in the Maxam-Gilbert DNA sequencing technique, chocolate and cocoa processing, soft drink processing, caramel coloring production and for thickening ice cream. It is often used to neutralize acids and prepare sodium salts of reagents. It forms a solution with sodium dodecyl sulfate and plays an important role for the decontamination of mycobacteria.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium hydroxide is widely used in variety of large scale applications such as pulp and paper manufacturing, in the production of soap and detergents as well as water treatment. It finds application in the base- driven hydrolysis of
TRUSTED_SUSTAINABILITY

Specifications

pH 14
Boiling Point 1390°C
CAS 1310-73-2
Melting Point 318°C
MDL Number MFCD00003548
Density 2.13
Solubility Information Soluble in water,methanol,glycerol and ethanol. Insoluble in acetone and ether.
Quantity 500 g
Formula Weight 40
Odor Odorless
Chemical Name or Material Sodium hydroxide
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0461565 Certificate TypeCertificate of Analysis Date15/10/2024 Catalog NumberA16037.36
Lot NumberA0458318 Certificate TypeCertificate of Analysis Date24/01/2024 Catalog NumberA16037.36
Lot NumberZ28J027 Certificate TypeCertificate of Analysis Date22/01/2024 Catalog NumberA16037.36
Lot NumberA0451178 Certificate TypeCertificate of Analysis Date09/01/2024 Catalog NumberA16037.36
Lot NumberW08J029 Certificate TypeCertificate of Analysis Date05/01/2024 Catalog NumberA16037.36
compliance-icons
Product Identifier
  • Sodium hydroxide, pellets
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : sodium hydroxide, sodium carbonate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Sodium hydroxide, pellets, 98% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.