Learn More
Sodium hydroxide, 97+%, ACS reagent, pellets, Thermo Scientific Chemicals
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Color | White |
Boiling Point | 1390.0°C |
Melting Point | 318.0°C |
Assay Percent Range | 97+% |
Linear Formula | NaOH |
Fieser | 01,1083; 05,616; 07,336; 08,460 |
Merck Index | 15, 8761 |
Solubility Information | Solubility in water: completely soluble. Other solubilities: soluble in alcohol and glycerol,insoluble in acetone |
Quantity | 1 kg |
Molecular Weight (g/mol) | 40 |
Show More |
Product Suggestions
Customers who viewed this item also viewed.
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
A0456762 | Certificate of Analysis | 09/04/2025 | 424330250,424330010,424330251,424335000,424330100,424330025 |
A0453016 | Certificate of Analysis | 25/01/2025 | 424330250,424330010,424335000,424330251,424330100,424330025 |
A0466917 | Certificate of Analysis | 21/10/2024 | 424330250,424330010,424330251,424335000,424330100,424330025 |
A0466918 | Certificate of Analysis | 21/10/2024 | 424330250,424330010,424330251,424335000,424330100,424330025 |
A0466916 | Certificate of Analysis | 21/10/2024 | 424330250,424330010,424330251,424335000,424330100,424330025 |
Safety and Handling

- Sodium hydroxide
- Danger
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 1 A
- Serious eye damage/eye irritation Category 1
- H290-May be corrosive to metals.
- H314-Causes severe skin burns and eye damage.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
Special Offers
-
Multiple Manufacturers
Chromatography and Chemicals Promotion
Explore our new promotion and get up to 70% off some of our most popular products.Promo Code: 24127Expires: 2025-05-31
Days
48Hours
10Minutes
19Seconds
19New Offer Save Up to 70% -
Multiple Manufacturers
Back to the Lab Summer Semester
Save up to 94% on Teaching Lab EssentialsPromo Code: 25721Expires: 2025-05-01
New Offer Save Now
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.