missing translation for 'onlineSavingsMsg'
Learn More

Potassium Iodate, Honeywell Fluka™

Puriss. p.a., 99.7-100.4%

£38.64 - £3955.65

Chemical Identifiers

CAS 7758-05-6
Molecular Formula IKO3
Molecular Weight (g/mol) 214.00
MDL Number MFCD00011406
InChI Key JLKDVMWYMMLWTI-UHFFFAOYSA-M
Synonym potassium iodate, potassium triodate, caswell no. 693a, iodic acid, potassium salt, unii-i139e44nhl, potassium iodine oxide kio3, iodic acid hio3 , potassium salt, epa pesticide chemical code 075703, iodic acid hio3 , potassium salt 1:1, kaliumjodat
PubChem CID 23665710
IUPAC Name potassium;iodate
SMILES [O-]I(=O)=O.[K+]
View More Specs

Products 4
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15682990
View Documents
Honeywell-Fluka
30314-100G
100 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15603000
View Documents
Honeywell-Fluka
30314-250G
250 g
£107.73
250g
Please to purchase this item. Need a web account? Register with us today!
15692990
View Documents
Honeywell-Fluka
30314-1KG
1 kg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15613000
View Documents
Honeywell-Fluka
30314-25KG
25 kg
£3955.65
25kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications

Chemical Identifiers

7758-05-6
214.00
JLKDVMWYMMLWTI-UHFFFAOYSA-M
23665710
[O-]I(=O)=O.[K+]
IKO3
MFCD00011406
potassium iodate, potassium triodate, caswell no. 693a, iodic acid, potassium salt, unii-i139e44nhl, potassium iodine oxide kio3, iodic acid hio3 , potassium salt, epa pesticide chemical code 075703, iodic acid hio3 , potassium salt 1:1, kaliumjodat
potassium;iodate

Specifications

7758-05-6
KIO3
1479
JLKDVMWYMMLWTI-UHFFFAOYSA-M
potassium;iodate
23665710
99.7 to 104%
3.93g/mL at 25°C
IKO3
MFCD00011406
potassium iodate, potassium triodate, caswell no. 693a, iodic acid, potassium salt, unii-i139e44nhl, potassium iodine oxide kio3, iodic acid hio3 , potassium salt, epa pesticide chemical code 075703, iodic acid hio3 , potassium salt 1:1, kaliumjodat
[O-]I(=O)=O.[K+]
214.00
214g/mol
puriss. p.a.
Potassium Iodate
Videos
Safety and Handling

Safety and Handling

P220-P261-P305 + P351 + P338

H272-H315-H319-H335

missing translation for 'einecsNumber' : 231-831-9

missing translation for 'rtecsNumber' : NN1350000

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.