Learn More
Potassium iodide, ACS, 99.0% min, Thermo Scientific Chemicals
Description
It is both a source of iodide and a mild reducing agent. It is used as a component in the electrolyte of dye sensitized solar cells. In the field of synthetic organic chemistry, it is used in the preparation of alkyl or aryl iodo compounds. It is employed as a nucleophilic catalyst in the alkylations with alkyl bromide, chloride, tosylate and mesylate. In the presence of acid it forms hydroiodic acid, which is a strong reducing agent. It is useful in producing Lugol’s solution (or Lugol’s iodine) which finds many applications including starch testing, gram stains, and etching of gold surfaces.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Boiling Point | 1330°C |
Melting Point | 681°C |
Color | White |
Physical Form | Granules |
Quantity | 500 g |
Assay Percent Range | 99.0% min |
Solubility Information | Soluble in water,alcohol,glycerol,and acetone. Slightly soluble in ether,and ammonia. |
Formula Weight | 166.01 |
Grade | ACS |
Odor | Odorless |
Show More |
Product Suggestions
Customers who viewed this item also viewed.
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
N06L004 | Certificate of Analysis | 10/02/2025 | 011601.22,011601.A3,011601.36 |
M21L044 | Certificate of Analysis | 23/01/2025 | 011601.22,011601.A3,011601.36 |
Z02K027 | Certificate of Analysis | 04/12/2024 | 011601.22,011601.A3,011601.36 |
X17K032 | Certificate of Analysis | 30/10/2024 | 011601.22,011601.A3,011601.36 |
W05K001 | Certificate of Analysis | 16/09/2024 | 011601.22,011601.A3,011601.36 |
Safety and Handling

- Potassium iodide
- Danger
- Specific target organ toxicity Category 1
- H372-Causes damage to organs.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P314-Get medical advice/attention if you feel unwell.
- MIXTURE LIST-Contains : Potassium iodide
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.