missing translation for 'onlineSavingsMsg'
Learn More
Potassium iodide, ACS, 99.0% min, Thermo Scientific Chemicals Product Code.: 11367347

Potassium iodide, ACS, 99.0% min, Thermo Scientific Chemicals

Product Code. 11367347
2 kg
Click to view available options
Quantity:
100 g
500 g
2 kg
This item is not returnable. View return policy

Product Code. 11367347

Brand: Thermo Scientific Alfa Aesar 011601.A3

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

For Use in Photographic Emulsions, in Analytical Chemistry, As a Feed Additive

It is both a source of iodide and a mild reducing agent. It is used as a component in the electrolyte of dye sensitized solar cells. In the field of synthetic organic chemistry, it is used in the preparation of alkyl or aryl iodo compounds. It is employed as a nucleophilic catalyst in the alkylations with alkyl bromide, chloride, tosylate and mesylate. In the presence of acid it forms hydroiodic acid, which is a strong reducing agent. It is useful in producing Lugol’s solution (or Lugol’s iodine) which finds many applications including starch testing, gram stains, and etching of gold surfaces.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7681-11-0
Molecular Formula IK
Molecular Weight (g/mol) 166.003
MDL Number MFCD00011405
InChI Key NLKNQRATVPKPDG-UHFFFAOYSA-M
Synonym potassium iodide, potassium iodide ki, pima, kali iodide, thyro-block, knollide, kisol, asmofug e, potassium diiodide, potassium monoiodideShow MoreShow Less
PubChem CID 4875
ChEBI CHEBI:8346
IUPAC Name potassium;iodide
SMILES [K+].[I-]

Specifications

Boiling Point 1330°C
Melting Point 681°C
Physical Form Granules
Quantity 2 kg
Merck Index 14,7643
Solubility Information Soluble in water,alcohol,glycerol,and acetone; Slightly soluble in ether,and ammonia.
Formula Weight 166.01
Percent Purity ≥99.0%
Grade ACS
Odor Odorless
Refractive Index 1.677
Sensitivity Hygroscopic
Density 3.12
Chemical Name or Material Potassium iodide
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberN06L004 Certificate TypeCertificate of Analysis Date10/02/2025 Catalog Number011601.22,011601.A3,011601.36
Lot NumberM21L044 Certificate TypeCertificate of Analysis Date23/01/2025 Catalog Number011601.22,011601.A3,011601.36
Lot NumberZ02K027 Certificate TypeCertificate of Analysis Date04/12/2024 Catalog Number011601.22,011601.A3,011601.36
Lot NumberX17K032 Certificate TypeCertificate of Analysis Date30/10/2024 Catalog Number011601.22,011601.A3,011601.36
Lot NumberW05K001 Certificate TypeCertificate of Analysis Date16/09/2024 Catalog Number011601.22,011601.A3,011601.36

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Potassium iodide, ACS, 99.0% min, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.