missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chemical Identifiers
CAS | 7681-11-0 |
---|---|
Molecular Formula | IK |
Molecular Weight (g/mol) | 166.003 |
MDL Number | 11405 |
InChI Key | NLKNQRATVPKPDG-UHFFFAOYSA-M |
Synonym | potassium iodide, potassium iodide ki, pima, kali iodide, thyro-block, knollide, kisol, asmofug e, potassium diiodide, potassium monoiodideShow More |
PubChem CID | 4875 |
ChEBI | CHEBI:8346 |
IUPAC Name | potassium;iodide |
SMILES | [K+].[I-] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11468772
|
Fisher Chemical
P/5840/48 |
100 g |
£104.00
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10766491
|
Fisher Chemical
P/5840/50 |
250 g |
£127.00
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10489180
|
Fisher Chemical
P/5840/53 |
500 g |
£259.00
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10132060
|
Fisher Chemical
P/5840/60 |
1 kg |
£437.00
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10409570
|
Fisher Chemical
P/5840/65 |
5 kg |
£1281.45
5kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Potassium Iodide, Extra Pure, SLR
Chemical Identifiers
7681-11-0 | |
166.003 | |
NLKNQRATVPKPDG-UHFFFAOYSA-M | |
4875 | |
potassium;iodide |
IK | |
11405 | |
potassium iodide, potassium iodide ki, pima, kali iodide, thyro-block, knollide, kisol, asmofug e, potassium diiodide, potassium monoiodideShow More | |
CHEBI:8346 | |
[K+].[I-] |
Specifications
1330°C | |
7681-11-0 | |
6 to 8 | |
IK | |
potassium iodide, potassium iodide ki, pima, kali iodide, thyro-block, knollide, kisol, asmofug e, potassium diiodide, potassium monoiodide | |
[K+].[I-] | |
166.003 | |
CHEBI:8346 | |
Plastic powder jar |
680°C | |
White | |
Solid | |
11405 | |
NLKNQRATVPKPDG-UHFFFAOYSA-M | |
potassium;iodide | |
4875 | |
166g/mol | |
1mmHg 745 |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Potassium iodide
Signal Word
- Danger
Hazard Category
- Specific target organ toxicity Category 1
Hazard Statement
- H372-Causes damage to organs.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P314-Get medical advice/attention if you feel unwell.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Potassium Iodide, Extra Pure, SLR, Fisher Chemical™