Learn More
Potassium nitrate, 99+%, ACS reagent, Thermo Scientific™
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications
Specifications
Quantity | 500g |
Assay Percent Range | 99+% |
Linear Formula | KNO3 |
UN Number | 1486 |
Fieser | 05,561 |
Merck Index | 15, 7769 |
Solubility Information | Solubility in water: 320g/L (20°C). Other solubilities: soluble in glycerol,slightly soluble in alcohol |
Formula Weight | 101.1 |
Percent Purity | 99+% |
Grade | ACS Reagent |
Show More |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
A0464950 | Certificate of Analysis | 12/08/2024 | 424155000,424150025,424151000 |
A0461411 | Certificate of Analysis | 04/04/2024 | 424155000,424150025,424151000 |
A0457817 | Certificate of Analysis | 15/11/2023 | 424155000,424150025,424151000 |
A0453512 | Certificate of Analysis | 25/08/2023 | 424155000,424150025,424151000 |
A0449330 | Certificate of Analysis | 03/02/2023 | 424155000,424150025,424151000 |
Safety and Handling

- Potassium nitrate
- Warning
- Oxidising solid Category 3
- H272-May intensify fire; oxidiser.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P220-Keep/Store away from clothing/combustible materials.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.