missing translation for 'onlineSavingsMsg'
Learn More
Potassium nitrate, 99+%, ACS reagent, Thermo Scientific™ Product Code.: 10023353

Potassium nitrate, 99+%, ACS reagent, Thermo Scientific™

Product Code. 10023353
100g, Glass bottle
Click to view available options
Quantity:
100g
500g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10023353

Brand: Thermo Scientific Acros 424151000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7757-79-1
Molecular Formula KNO3
Molecular Weight (g/mol) 101.1
MDL Number MFCD00011409
InChI Key FGIUAXJPYTZDNR-UHFFFAOYSA-N
Synonym potassium nitrate, saltpeter, nitre, nitrate of potash, vicknite, nitric acid potassium salt, nitric acid, potassium salt, saltpetre, kaliumnitrat, kalii nitrasShow MoreShow Less
PubChem CID 24434
ChEBI CHEBI:63043
IUPAC Name potassium;nitrate
SMILES [N+](=O)([O-])[O-].[K+]

Specifications

Quantity 100g
Assay Percent Range 99+%
Linear Formula KNO3
UN Number 1486
Fieser 05,561
Merck Index 15, 7769
Solubility Information Solubility in water: 320g/L (20°C). Other solubilities: soluble in glycerol,slightly soluble in alcohol
Formula Weight 101.1
Percent Purity 99+%
Grade ACS Reagent
Packaging Glass bottle
Density 2.1
Chemical Name or Material Potassium nitrate, crystals
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0464950 Certificate TypeCertificate of Analysis Date12/08/2024 Catalog Number424155000,424150025,424151000
Lot NumberA0461411 Certificate TypeCertificate of Analysis Date04/04/2024 Catalog Number424155000,424150025,424151000
Lot NumberA0457817 Certificate TypeCertificate of Analysis Date15/11/2023 Catalog Number424155000,424150025,424151000
Lot NumberA0453512 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number424155000,424150025,424151000
Lot NumberA0449330 Certificate TypeCertificate of Analysis Date03/02/2023 Catalog Number424155000,424150025,424151000
compliance-icons
Product Identifier
  • Potassium nitrate
Signal Word
  • Warning
Hazard Category
  • Oxidising solid Category 3
Hazard Statement
  • H272-May intensify fire; oxidiser.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P220-Keep/Store away from clothing/combustible materials.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Potassium nitrate, 99+%, ACS reagent, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.