missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium nitrate, ACS, 99.0% min, Thermo Scientific™
Click to view available options
Quantity:
100g
500g
2kg
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications
Specifications
Quantity | 100g |
Assay Percent Range | 99.0% min. |
Formula Weight | 101.11 |
Grade | ACS |
Odor | Odorless |
Appearance | White crystalline |
Density | 2.109g/cm3 |
Chemical Name or Material | Potassium nitrate, For ACS analysis |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
62500199 | Certificate of Analysis | 11/04/2025 | 013443.22,013443.A3,013443.36 |
N28L050 | Certificate of Analysis | 05/03/2025 | 013443.22,013443.A3,013443.36 |
N25L011 | Certificate of Analysis | 27/02/2025 | 013443.22,013443.A3,013443.36 |
108931 | Certificate of Analysis | 12/02/2025 | 013443.22,013443.A3,013443.36 |
N03L013 | Certificate of Analysis | 05/02/2025 | 013443.22,013443.A3,013443.36 |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Potassium nitrate, ACS, 99.0% min, Thermo Scientific™ >
Spot an opportunity for improvement?Share a Content Correction