missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Oxalate Monohydrate, Certified AR for Analysis, Fisher Chemical™
£103.00 - £263.00
Chemical Identifiers
CAS | 6487-48-5 |
---|---|
Molecular Formula | C2H2K2O5 |
Molecular Weight (g/mol) | 184.23 |
MDL Number | MFCD00150033 |
InChI Key | QCPTVXCMROGZOL-UHFFFAOYSA-L |
Synonym | potassium oxalate monohydrate, potassium oxalate hydrate, oxalic acid potassium salt, ethanedioic acid, dipotassium salt, monohydrate, dipotassium oxalate hydrate, dipotassium hydrate oxalate, acmc-1bcm6, ksc495a4h, dipotassium oxalate monohydrate, dipotassium ethanedioate hydrate |
PubChem CID | 2724193 |
SMILES | O.[K+].[K+].[O-]C(=O)C([O-])=O |
Description
Chemical Identifiers
6487-48-5 | |
184.23 | |
QCPTVXCMROGZOL-UHFFFAOYSA-L | |
2724193 |
C2H2K2O5 | |
MFCD00150033 | |
potassium oxalate monohydrate, potassium oxalate hydrate, oxalic acid potassium salt, ethanedioic acid, dipotassium salt, monohydrate, dipotassium oxalate hydrate, dipotassium hydrate oxalate, acmc-1bcm6, ksc495a4h, dipotassium oxalate monohydrate, dipotassium ethanedioate hydrate | |
O.[K+].[K+].[O-]C(=O)C([O-])=O |
Specifications
356°C | |
White | |
Solid | |
MFCD00150033 | |
QCPTVXCMROGZOL-UHFFFAOYSA-L | |
dipotassium;oxalate;hydrate | |
2724193 | |
Plastic powder jar |
6487-48-5 | |
7.0 to 8.5 | |
C2H2K2O5 | |
potassium oxalate monohydrate, potassium oxalate hydrate, oxalic acid potassium salt, ethanedioic acid, dipotassium salt, monohydrate, dipotassium oxalate hydrate, dipotassium hydrate oxalate, acmc-1bcm6, ksc495a4h, dipotassium oxalate monohydrate, dipotassium ethanedioate hydrate | |
O.[K+].[K+].[O-]C(=O)C([O-])=O | |
184.23 | |
184.24g/mol |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Potassium oxalate monohydrate
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
Hazard Statement
- H312-Harmful in contact with skin.
- H302-Harmful if swallowed.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
Supplemental information
- MIXTURE LIST-Contains : Oxalate, potassium,monohydrate, Potassium oxalate