missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Permanganate, Extra Pure, SLR, Fisher Chemical™
KMnO4, CAS Number-7722-64-7, solo san soo, chameleon mineral, cairox, walko tablets, argucide, potassium permanganate, insta-perm, algae-k, condy's crystals, permanganate of potash, 500g, Brown, 158.04g/mol, VZJVWSHVAAUDKD-UHFFFAOYSA-N, potassium;permanganate, 240 deg.C, 11364, 158.032, Glass Jar
£50.50 - £247.00
Chemical Identifiers
CAS | 7722-64-7 |
---|---|
Molecular Formula | KMnO4 |
Molecular Weight (g/mol) | 158.032 |
MDL Number | 11364 |
InChI Key | VZJVWSHVAAUDKD-UHFFFAOYSA-N |
Synonym | potassium permanganate, chameleon mineral, condy's crystals, argucide, cairox, permanganate of potash, insta-perm, walko tablets, algae-k, solo san soo |
PubChem CID | 516875 |
IUPAC Name | potassium;permanganate |
SMILES | [O-][Mn](=O)(=O)=O.[K+] |
Description
KMnO4, CAS Number-7722-64-7, solo san soo, chameleon mineral, cairox, walko tablets, argucide, potassium permanganate, insta-perm, algae-k, condy's crystals, permanganate of potash, 500g, Brown, 158.04g/mol, VZJVWSHVAAUDKD-UHFFFAOYSA-N, potassium;permanganate, 240 deg.C, 11364, 158.032, Glass Jar
Chemical Identifiers
7722-64-7 | |
158.032 | |
VZJVWSHVAAUDKD-UHFFFAOYSA-N | |
516875 | |
[O-][Mn](=O)(=O)=O.[K+] |
KMnO4 | |
11364 | |
potassium permanganate, chameleon mineral, condy's crystals, argucide, cairox, permanganate of potash, insta-perm, walko tablets, algae-k, solo san soo | |
potassium;permanganate |
Specifications
240°C | |
Brown | |
Powder Solid | |
11364 | |
VZJVWSHVAAUDKD-UHFFFAOYSA-N | |
potassium;permanganate | |
516875 | |
Extra Pure |
7722-64-7 | |
8 | |
KMnO4 | |
potassium permanganate, chameleon mineral, condy's crystals, argucide, cairox, permanganate of potash, insta-perm, walko tablets, algae-k, solo san soo | |
[O-][Mn](=O)(=O)=O.[K+] | |
158.032 | |
158.04g/mol |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Potassium permanganate
Signal Word
- Danger
Hazard Category
- Oxidising solid Category 2
- Acute toxicity Category 4
- Skin corrosion/irritation Category 1 C
- Serious eye damage/eye irritation Category 1
- Specific target organ toxicity after repeated exposure Category 2
- ACUTE AQUATIC Acute 1
- LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
- H272-May intensify fire; oxidiser.
- H302-Harmful if swallowed.
- H314-Causes severe skin burns and eye damage.
- H373-May cause damage to organs.
- H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Potassium Permanganate, Extra Pure, SLR, Fisher Chemical™