missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Phosphate, Dibasic, 1.0M solution in water, Thermo Scientific Chemicals
Brand: Thermo Scientific Acros 397080025
55.65 GBP valid until 2025-01-31
Use promo code "23894" to get your promotional price.
This item is not returnable.
View return policy
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Potassium phosphate, dibasic | |
2.5 L | |
K2HPO4 | |
15, 7779 | |
ZPWVASYFFYYZEW-UHFFFAOYSA-L | |
dipotassium;hydrogen phosphate | |
24450 | |
174.18 | |
Glass bottle | |
1.13 |
7758-11-4, 7732-18-5 | |
HK2O4P | |
MFCD00011383 | |
dipotassium hydrogen phosphate, dipotassium phosphate, dipotassium hydrogenphosphate, dibasic potassium phosphate, potassium hydrogen phosphate, potassium phosphate, dibasic, potassium dibasic phosphate, potassium phosphate dibasic, phosphoric acid, dipotassium salt, dipotassium monophosphate | |
OP(=O)([O-])[O-].[K+].[K+] | |
174.18 | |
CHEBI:32031 | |
1.0M, exact strength on the certificate of analysis | |
Clear colorless solution | |
1.1300g/mL |
RUO – Research Use Only