missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Phosphate, Dibasic, 1.0M solution in water, Thermo Scientific Chemicals
£31.10 - £72.90
Specifications
Chemical Name or Material | Potassium phosphate, dibasic |
---|---|
CAS | 7758-11-4, 7732-18-5 |
Molecular Formula | HK2O4P |
Linear Formula | K2HPO4 |
MDL Number | MFCD00011383 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10173064
|
Thermo Scientific Acros
397080010 |
1 L | Glass bottle |
£31.10
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10729005
|
Thermo Scientific Acros
397080025 |
2.5 L | Glass bottle |
£72.90
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Potassium phosphate, dibasic | |
HK2O4P | |
MFCD00011383 | |
dipotassium hydrogen phosphate, dipotassium phosphate, dipotassium hydrogenphosphate, dibasic potassium phosphate, potassium hydrogen phosphate, potassium phosphate, dibasic, potassium dibasic phosphate, potassium phosphate dibasic, phosphoric acid, dipotassium salt, dipotassium monophosphate | |
OP(=O)([O-])[O-].[K+].[K+] | |
174.18 | |
CHEBI:32031 | |
1.0M, exact strength on the certificate of analysis | |
Clear colorless solution | |
1.1300g/mL |
7758-11-4, 7732-18-5 | |
K2HPO4 | |
15, 7779 | |
ZPWVASYFFYYZEW-UHFFFAOYSA-L | |
dipotassium;hydrogen phosphate | |
24450 | |
174.18 | |
Glass bottle | |
1.13 |
Safety and Handling
missing translation for 'einecsNumber' : 231-834-5
RUO – Research Use Only