missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Phosphate NF GenAR Crystal, Macron Fine Chemicals™
macron
£148.00 - £525.00
Chemical Identifiers
CAS | 7778-77-0 |
---|---|
Molecular Formula | H2KO4P |
Molecular Weight (g/mol) | 136.08 |
MDL Number | MFCD00011401 MFCD00147253 |
InChI Key | GNSKLFRGEWLPPA-UHFFFAOYSA-M |
Synonym | potassium dihydrogen phosphate, monopotassium phosphate, potassium phosphate monobasic, potassium phosphate, monobasic, potassium acid phosphate, phosphoric acid, monopotassium salt, monobasic potassium phosphate, monopotassium monophosphate, monopotassium dihydrogen phosphate, monopotassium orthophosphate |
PubChem CID | 516951 |
ChEBI | CHEBI:63036 |
IUPAC Name | potassium dihydrogen phosphate |
SMILES | [K+].OP(O)([O-])=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
15578434
|
Avantor Macron
7746-04 |
500 g | POLYSTORMOR |
£148.00
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15588434
|
Avantor Macron
7746-06 |
2.5 kg | Poly |
£525.00
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Chemical Identifiers
7778-77-0 | |
136.08 | |
GNSKLFRGEWLPPA-UHFFFAOYSA-M | |
516951 | |
potassium dihydrogen phosphate |
H2KO4P | |
MFCD00011401 MFCD00147253 | |
potassium dihydrogen phosphate, monopotassium phosphate, potassium phosphate monobasic, potassium phosphate, monobasic, potassium acid phosphate, phosphoric acid, monopotassium salt, monobasic potassium phosphate, monopotassium monophosphate, monopotassium dihydrogen phosphate, monopotassium orthophosphate | |
CHEBI:63036 | |
[K+].OP(O)([O-])=O |
Specifications
7778-77-0 | |
H2KO4P | |
potassium dihydrogen phosphate, monopotassium phosphate, potassium phosphate monobasic, potassium phosphate, monobasic, potassium acid phosphate, phosphoric acid, monopotassium salt, monobasic potassium phosphate, monopotassium monophosphate, monopotassium dihydrogen phosphate, monopotassium orthophosphate | |
[K+].OP(O)([O-])=O | |
136.08 | |
CHEBI:63036 | |
NF |
Crystalline | |
MFCD00011401 MFCD00147253 | |
GNSKLFRGEWLPPA-UHFFFAOYSA-M | |
potassium dihydrogen phosphate | |
516951 | |
136.09 | |
Potassium Phosphate, Monobasic |