Learn More
Potassium phosphate, tribasic monohydrate, 96%, extra pure, Thermo Scientific Chemicals
£16.00 - £1155.00
Chemical Identifiers
CAS | 27176-10-9 |
---|---|
Molecular Formula | K3O4P·H2O |
Molecular Weight (g/mol) | 230.28 |
MDL Number | MFCD00150410 |
InChI Key | RMNIZOOYFMNEJJ-UHFFFAOYSA-K |
Synonym | tri-potassium phosphate monohydrate, potassium phosphate tribasic monohydrate, potassium phosphate hydrate, potassium phosphate monohydrate, unii-4927m96tfv, tert-potassium phosphate monohydrate, potassium phosphate, tribasic, monohydrate, phosphoric acid, tripotassium salt, monohydrate, tripotassium phosphate, monohydrate, acmc-1chx7 |
PubChem CID | 22031701 |
IUPAC Name | tripotassium;phosphate;hydrate |
SMILES | O.[O-]P(=O)([O-])[O-].[K+].[K+].[K+] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10751331
|
Thermo Scientific Acros
263790250 |
25 g | Glass bottle |
£16.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10048521
|
Thermo Scientific Acros
263795000 |
500 g | Glass bottle |
£30.40
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10003340
|
Thermo Scientific Acros
263790010 |
1 kg | Glass bottle |
£50.30
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10511984
|
Thermo Scientific Acros
263790050 |
5 kg | Plastic Bottle |
£212.00
5kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10267123
|
Thermo Scientific Acros
263790251 |
25 kg | Fibre drum |
£1155.00
25kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
27176-10-9 | |
230.28 | |
RMNIZOOYFMNEJJ-UHFFFAOYSA-K | |
22031701 | |
O.[O-]P(=O)([O-])[O-].[K+].[K+].[K+] |
K3O4P·H2O | |
MFCD00150410 | |
tri-potassium phosphate monohydrate, potassium phosphate tribasic monohydrate, potassium phosphate hydrate, potassium phosphate monohydrate, unii-4927m96tfv, tert-potassium phosphate monohydrate, potassium phosphate, tribasic, monohydrate, phosphoric acid, tripotassium salt, monohydrate, tripotassium phosphate, monohydrate, acmc-1chx7 | |
tripotassium;phosphate;hydrate |
Specifications
1340.0°C | |
White | |
96% | |
K3PO4·H2O | |
tri-potassium phosphate monohydrate, potassium phosphate tribasic monohydrate, potassium phosphate hydrate, potassium phosphate monohydrate, unii-4927m96tfv, tert-potassium phosphate monohydrate, potassium phosphate, tribasic, monohydrate, phosphoric acid, tripotassium salt, monohydrate, tripotassium phosphate, monohydrate, acmc-1chx7 | |
RMNIZOOYFMNEJJ-UHFFFAOYSA-K | |
tripotassium;phosphate;hydrate | |
22031701 | |
96% | |
13% max. (800°C) | |
Potassium phosphate, tribasic monohydrate, c.p. |
27176-10-9 | |
Powder | |
K3O4P·H2O | |
MFCD00150410 | |
Solubility in water: soluble. Other solubilities: insoluble in alcohol | |
O.[O-]P(=O)([O-])[O-].[K+].[K+].[K+] | |
230.28 | |
230.28 | |
Extra Pure | |
2.564 |
Safety and Handling
GHS H Statement
Causes serious eye damage.
May cause respiratory irritation.
GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mis
GHS Signal Word: Danger
- Potassium phosphate tribasic monohydrate
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 1
- Specific target organ toxicity Category 3
Hazard Statement
- H335-May cause respiratory irritation.
- H318-Causes serious eye damage.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only