Learn More
Potassium selenocyanate, 98.5%, Thermo Scientific Chemicals
Description
Potassium selenocyanate is used to prepare triphenylphosphine selenide by reacting with triphenylphosphine. It finds application in solid state NMR of heavy-metal nuclei which results in the preparation of several mercury thiocyanate and selenocyanate compounds.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | 162°C to 164°C |
Color | White |
Physical Form | Crystalline |
Quantity | 50 g |
UN Number | UN3283 |
Solubility Information | Soluble in water,ethanol,dimethyl formamide,hexamethylphosphoramide,acetonitrile and methanol. Slightly soluble in terahydrofuran. |
Formula Weight | 144.08 |
Percent Purity | 98.5% |
Odor | Odorless |
Sensitivity | Moisture sensitive |
Show More |
Product Suggestions
Customers who viewed this item also viewed.
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
M30L002 | Certificate of Analysis | 03/02/2025 | 068107.18,068107.09 |
M30L001 | Certificate of Analysis | 03/02/2025 | 068107.18,068107.09 |
X27H046 | Certificate of Analysis | 12/04/2022 | 068107.18,068107.09 |
P01I014 | Certificate of Analysis | 12/04/2022 | 068107.18,068107.09 |
P10G001 | Certificate of Analysis | 12/04/2022 | 068107.18,068107.09 |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.