missing translation for 'onlineSavingsMsg'
Learn More

Potassium sodium tartrate tetrahydrate, 99+%, for analysis, Thermo Scientific Chemicals

£46.53 - £947.72

Chemical Identifiers

CAS 6381-59-5
MDL Number MFCD00150989
InChI Key VZOPRCCTKLAGPN-UHFFFAOYSA-L
Synonym potassium sodium tartrate tetrahydrate, sodium potassium tartrate tetrahydrate, monopotassium monosodium tartrate tetrahydrate, sodiumpotassiumtartrate,tetr, potassium sodium 2,3-dihydroxybutanedioate tetrahydrate, potassium sodium tetrahydrate tartrate, l +-tartaric acid potassium sodium salt, acmc-209nif, acmc-20aju8, ksc493o7j
PubChem CID 2724148
IUPAC Name potassium;sodium;2,3-dihydroxybutanedioate;tetrahydrate
SMILES C(C(C(=O)[O-])O)(C(=O)[O-])O.O.O.O.O.[Na+].[K+]
View More Specs

Products 4
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10462011
View Documents Promotion Details
Thermo Scientific Acros
202865000
500 g Plastic bottle
£46.53
500g
Please to purchase this item. Need a web account? Register with us today!
10721861
View Documents
Thermo Scientific Acros
202860010
1 kg Plastic bottle
£77.49
1kg
Please to purchase this item. Need a web account? Register with us today!
10638343
View Documents
Thermo Scientific Acros
202860051
5 kg Plastic Drum
£261.30
5kg
Please to purchase this item. Need a web account? Register with us today!
15109233
View Documents
Thermo Scientific Acros
202860250
25 kg Fibre drum
£947.72
25kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

6381-59-5
VZOPRCCTKLAGPN-UHFFFAOYSA-L
2724148
C(C(C(=O)[O-])O)(C(=O)[O-])O.O.O.O.O.[Na+].[K+]
MFCD00150989
potassium sodium tartrate tetrahydrate, sodium potassium tartrate tetrahydrate, monopotassium monosodium tartrate tetrahydrate, sodiumpotassiumtartrate,tetr, potassium sodium 2,3-dihydroxybutanedioate tetrahydrate, potassium sodium tetrahydrate tartrate, l +-tartaric acid potassium sodium salt, acmc-209nif, acmc-20aju8, ksc493o7j
potassium;sodium;2,3-dihydroxybutanedioate;tetrahydrate

Specifications

6381-59-5
White
Authentic
C4H12KNaO10
MFCD00150989
15, 7790
potassium sodium tartrate tetrahydrate, sodium potassium tartrate tetrahydrate, monopotassium monosodium tartrate tetrahydrate, sodiumpotassiumtartrate,tetr, potassium sodium 2,3-dihydroxybutanedioate tetrahydrate, potassium sodium tetrahydrate tartrate, l +-tartaric acid potassium sodium salt, acmc-209nif, acmc-20aju8, ksc493o7j
C(C(C(=O)[O-])O)(C(=O)[O-])O.O.O.O.O.[Na+].[K+]
282.218
282.22
Analysis
Potassium sodium tartrate tetrahydrate
>100°C
5.5 to 8.5 (5% soln. at 20°C)
99+%
KO2CCH(OH)CH(OH)CO2Na·4H2O
03, 525
+ 22.10 (20.00°C c=10,water)
VZOPRCCTKLAGPN-UHFFFAOYSA-L
potassium;sodium;2,3-dihydroxybutanedioate;tetrahydrate
2724148
99 to 102%
Crystals
Videos
SDS
Documents

Documents

Product Certifications

RUO – Research Use Only