Learn More
Heidolph Instruments Powder flask
For use with rotary evaporators
Brand: Heidolph Instruments 514-74200-00
This item is not returnable.
View return policy
Description
Powder flasks come with a standard NS 29/32 joint size but are also available with NS 24/29 ground joints.Specifications
Borosilicate Glass | |
Evaporating Flask | |
1000 mL | |
115/230 VAC |