missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Usbeck™ Powder Spatula
Transfer substances with simplicity. Usbeck™ Powder Spatula is ideal for transferring substances.
£1.90 - £2.20
Product Code | Brand | Length (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Length (Metric) | Price | Quantity & Availability | |||||
10691442
|
Usbeck™
3282 |
150 mm |
£1.90
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10760503
|
Usbeck™
3284 |
210 mm |
£2.20
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- Stainless steel
- Width of blade 9 mm
Specifications
Powder Spatula | |
Stainless Steel |