Learn More
Invitrogen™ Power Blotter Pre-cut Membranes and Filters, nitrocellulose, regular size
Precut, preassembled nitrocellulose membrane and filter paper sandwiches for gels
£253.00
Specifications
Material Nitrocellulose
Shipping Condition Room Temperature
View More Specs
Products 1
Product Code Brand Price Quantity & Availability  
Product Code Brand Price Quantity & Availability  
15858502
missing translation for 'viewDocuments' Promotion Details
Invitrogen™
PB7320
Description: Fits standard, precast or homemade midi gels (13cm x 8cm) or two mini gels; Size: 0.2µM pore size
£253.00
Pack of 20
View alternative products
missing translation for 'toSaveUpto' 
»
In Stock
to see stock.
Add to basket
Description
Description
The Invitrogen Power Blotter Pre-cut Membranes and Filters are designed for transferring proteins from gels using the Invitrogen Power Blotter System.
Features:
  • Membranes and filters allow for high efficiency semi-dry transfer of proteins in less than 10 minutes when combined with the Power Blotter 1-Step Transfer Buffer
  • 100% pure nitrocellulose membrane (0.2 μM pore size) with high surface area and excellent uniformity provides high-quality transfers.
    • Nitrocellulose is a popular binding matrix for western blotting because of its high affinity for proteins and compatibility with a variety of detection methods such as chemiluminescence, chromogenic, and fluorescence. 
    • 209 μg/cm2 protein binding affinity
    • Blocks easily produces very low background in chemiluminescent and fluorescent western blotting
  • Convenient— Pre-cut sheets are ready to use, meaning no need to cut your own membranes, minimizing damage and contamination
  • Validated – When combined, membrane and filter provide the thickness and buffer capacity needed for optimal performance
Specifications
Specifications
Nitrocellulose
Room Temperature