missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Power Blotter Select Transfer Stacks, nitrocellulose, mini

Transfer proteins rapidly and efficiently with these ready-to-use high performance transfer stacks for Invitrogen Power Blotter.

Brand:  Invitrogen™ PB3210

64.66 GBP valid until 2024-12-20
Use promo code "21651" to get your promotional price.


 View more versions of this product

Product Code. 15826162

  • £106.00 / Each
Estimated Shipment: 27-11-2024
to see stock.
Edge
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

The Invitrogen Power Blotter Select Transfer Stacks are used to transfer proteins using the Invitrogen Power Blotter System. Disposable, pre-assembled stacks come with an integrated nitrocellulose transfer membrane to perform dry blotting of proteins from polyacrylamide gels. Each Power Blotter Select Transfer Stack contains filter paper, membrane and the appropriate cathode and anode buffers incorporated into a solid gel matrix to allow fast, reliable transfer of proteins.

Nitrocellulose Membrane:

  • 0.2 μm pore size
  • Composed of 100% pure nitrocellulose for high-quality transfer.
  • Compatible with commonly used detection methods such as staining, chemiluminescence, or fluorescence.
  • Proteins bind as hydrophobic and electrostatic interactions.
  • Protein binding capacity is 209 μg/cm2.

Rapid, efficient and even transfer:

  • Ready-to-use transfer stack takes little time to set up
  • Protein transfers in as little as 7 minutes
  • Stacks made with high quality membranes for high binding efficiency, uniform transfer, and high detection sensitivity

Increased reproducibility:

  • Preassembled stacks provide consistent performance

 

TRUSTED_SUSTAINABILITY
Specifications

Specifications

8 cm
Transfer Stack
0.2 μm
Room Temperature
Power Blotter
8 cm
Nitrocellulose
10 mini-size transfer stacks (8 cm x 8 cm)
Mini, 8 cm x 8 cm
10 Blots
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.