Learn More
Invitrogen™ Power Blotter Select Transfer Stacks, nitrocellulose, regular size
Transfer proteins rapidly and efficiently with these ready-to-use high performance transfer stacks for Invitrogen Power Blotter.
£135.00 - £468.00
Specifications
Length (Metric) | 13 cm |
---|---|
Width (Metric) | 8.3 cm |
Format | Transfer Stack |
Material | Nitrocellulose |
Pore Size | 0.2 μm |
Product Code | Brand | Sufficient For | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Sufficient For | Price | Quantity & Availability | |||||
15846162
|
Invitrogen™
PB3310 |
10 Blots |
£135.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15856162
|
Invitrogen™
PB3340 |
40 Blots |
£468.00
Pack of 4 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The Invitrogen Power Blotter Select Transfer Stacks are used to transfer proteins using the Invitrogen Power Blotter System. Disposable, pre-assembled stacks come with an integrated nitrocellulose transfer membrane to perform dry blotting of proteins from polyacrylamide gels. Each Power Blotter Select Transfer Stack contains filter paper, membrane and the appropriate cathode and anode buffers incorporated into a solid gel matrix to allow fast, reliable transfer of proteins.
Nitrocellulose Membrane:
- 0.2 μm pore size
- Composed of 100% pure nitrocellulose for high-quality transfer.
- Compatible with commonly used detection methods such as staining, chemiluminescence, or fluorescence.
- Proteins bind as hydrophobic and electrostatic interactions.
- Protein binding capacity is 209 μg/cm2.
Rapid, efficient and even transfer:
- Ready-to-use transfer stack takes little time to set up
- Protein transfers in as little as 7 minutes
- Stacks made with high quality membranes for high binding efficiency, uniform transfer, and high detection sensitivity
Increased reproducibility:
- Preassembled stacks provide consistent performance
Specifications
13 cm | |
Transfer Stack | |
0.2 μm | |
Regular, 13 cm x 8.3 cm |
8.3 cm | |
Nitrocellulose | |
Room Temperature | |
Power Blotter |
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.