Learn More
Invitrogen™ Power Blotter Welcome Pack
Get rapid and efficient semi-dry protein transfer with the Invitrogen™ Power Blotter.
Brand: Invitrogen™ PB0112
2158.40 GBP valid until 2024-12-15
Use promo code "21658" to get your promotional price.
Includes: Power Blotter Station; Power Blotter Cassette; Power Blotter Select Transfer Stacks, Nitrocellulose, Regular; Power Blotter Pre-cut Membranes and Filters, Nitrocellulose, Regular; Power Blotter 1-Step Transfer Buffer
Description
The Invitrogen™ Power Blotter System is designed specifically for rapid semi-dry transfer of proteins from polyacrylamide gels to nitrocellulose or PVDF membranes in 5 to 12 minutes. The Power Blotter features an integrated power supply optimized to enable consistent and high-efficiency protein transfer.
The Power Blotter achieves highly efficient transfer in 7 minutes when used with Power Blotter Select Transfer Stacks or 10 minutes with the Power Blotter 1-Step Transfer Buffer. The instrument is also effective for standard 30- to 60-minute semi-dry transfer protocols based on traditional buffers.
Features:
- Efficient— high transfer efficiency with a broad range of protein sizes compared to conventional semi-dry or wet (tank) transfer methods
- Fast—5 to 12 minute transfer when used with Power Blotter Select Transfer Stacks or Power Blotter 1-Step Transfer Buffer
- Integrated power supply— seamless operation between control unit and cassette; no need for external power supply
- Easy-touch programming— convenient pre-programmed transfer methods or create, save, and run customized transfer methods
- Flexible— allows for simultaneous transfer
- Versatile—compatible with Power Blotter Select Transfer Stacks, Pierce 1-Step Transfer Buffer for rapid blotting programs or Towbin transfer buffer for conventional semi-dry transfer methods
Specifications
Novex Bolt gels, NuPage Protein gels | |
Nitrocellulose | |
Room Temperature | |
Power Blotter |
1 Kit | |
• Power Blotter Station • Power Blotter Cassette • Power Blotter Select Transfer Stacks, nitrocellulose, regular-size • Power Blotter Pre-cut Membranes and Filters, nitrocellulose, regular-size • Power Blotter 1-Step Transfer Buffer |
|
Semi-Dry |
For Research Use Only. Not for use in diagnostic procedures.