Learn More
Invitrogen™ Power Blotter XL Welcome Pack, PVDF
Designed specifically for rapid semi-dry transfer of proteins from polyacrylamide gels to nitrocellulose or PVDF membranes in 5 to 12 minutes
Brand: Invitrogen™ PB0117
2397.80 GBP valid until 2024-12-15
Use promo code "21658" to get your promotional price.
This item is not returnable.
View return policy
Description
- The Invitrogen Power Blotter XL System (Power Blotter Station + Power Blotter Cassette XL) is designed specifically for rapid semi-dry transfer of proteins from polyacrylamide gels to nitrocellulose or PVDF membranes in 5 to 12 minutes.
- The Power Blotter System features an integrated power supply optimized to enable consistent and high-efficiency protein transfer.
- The Power Blotter XL Welcome Pack, PVDF, includes: Power Blotter Station, Power Blotter Cassette XL, Power Blotter Select Transfer Stacks, PVDF, regular size, Power Blotter Pre-cut Membranes and Filters, PVDF, regular size, Power Blotter 1-Step Transfer Buffer
- Efficient—high transfer efficiency with a broad range of protein sizes compared to conventional semi-dry or wet (tank) transfer methods
- Fast—5- to 12-minute transfer when used with Power Blotter Select Transfer Stacks or Power Blotter 1-Step Transfer Buffer
- Integrated power supply—seamless operation between control unit and cassette; no need for external power supply
- Easy-touch programming—convenient pre-programmed transfer methods, or create, save, and run customized transfer methods
- Flexible—simultaneous transfer of 1 to 4 mini-size or 1 or 2 midi-size gels using the Power Blotter XL Cassette
- Versatile—compatible with Power Blotter Select Transfer Stacks and Pierce 1-Step Transfer Buffer for rapid blotting programs or Towbin transfer buffer for conventional semi-dry transfer methods
- The Power Blotter System achieves highly efficient transfer in seven minutes when used with Power Blotter Select Transfer Stacks or ten minutes with Power Blotter 1-Step Transfer Buffer. The instrument is also effective for standard 30- to 60-minute semi-dry transfer protocols based on traditional buffers.
Specifications
Acrylamide Gels | |
PVDF | |
Room Temperature | |
Power Blotter |
1 kit | |
• Power Blotter Station • Power Blotter Cassette XL • Power Blotter Select Transfer Stacks, PVDF, regular-size • Power Blotter Pre-cut Membranes and Filters, PVDF, regular-size • Power Blotter 1-Step Transfer Buffer |
|
Semi-Dry |
For Research Use Only. Not for use in diagnostic procedures.