Learn More
Brady™ PowerMark™ Vinyl Labelling Tape
Brady's B-595 vinyl will adhere to difficult low surface energy items, including: PVC piping, blow-molded equipment cases, ABS plastics, powder-coated surfaces and equipment made from recycled plastics. PowerMark™ Vinyl Labelling Tape also excels at sticking to highly textured surfaces.
£171.00 - £242.00
Specifications
Certifications/Compliance | RoHS compliant to 2005/618/EC MCV amendment to RoHS Directive 2002/95/EC |
---|---|
Adhesive Type | Permanent Acrylic, Permanent |
Finish | Gloss |
Format | Roll |
Material | Vinyl |
Product Code | Brand | Color | Dimensions (LxW) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Color | Dimensions (LxW) | Price | Quantity & Availability | ||||
11552695
|
Brady™
013605 |
Black | 15.24 m x 152.4 mm |
£205.00
1 box |
Estimated Shipment: 06-09-2024
Log in to see stock. |
||||
11502645
|
Brady™
013544 |
Black | 15.24 m x 101.6 mm |
£171.00
1 box |
Estimated Shipment: 06-09-2024
Log in to see stock. |
||||
11532655
|
Brady™
013557 |
Black | 15.24 m x 203.20 mm |
£231.00
1 box |
Estimated Shipment: 06-09-2024
Log in to see stock. |
||||
11542705
|
Brady™
013614 |
Black | 15.24 m x 254 mm |
£242.00
1 box |
Estimated Shipment: 08-08-2024
Log in to see stock. |
||||
11572635
|
Brady™
013541 |
Blue | 15.24 m x 101.6 mm |
£171.00
1 box |
Estimated Shipment: 08-08-2024
Log in to see stock. |
||||
11522695
|
Brady™
013602 |
Blue | 15.24 m x 152.4 mm |
£205.00
1 box |
Estimated Shipment: 08-08-2024
Log in to see stock. |
||||
11502655
|
Brady™
013554 |
Blue | 15.24 m x 203.20 mm |
£231.00
1 box |
Estimated Shipment: 06-09-2024
Log in to see stock. |
||||
11512705
|
Brady™
013611 |
Blue | 15.24 m x 254 mm |
£242.00
1 box |
Estimated Shipment: 08-08-2024
Log in to see stock. |
||||
11532645
|
Brady™
013547 |
Brown | 15.24 m x 101.6 mm |
£171.00
1 box |
Estimated Shipment: 06-09-2024
Log in to see stock. |
||||
11562655
|
Brady™
013560 |
Brown | 15.24 m x 203.20 mm |
£231.00
1 box |
Estimated Shipment: 06-09-2024
Log in to see stock. |
||||
11582695
|
Brady™
013608 |
Brown | 15.24 m x 152.4 mm |
£205.00
1 box |
Estimated Shipment: 08-08-2024
Log in to see stock. |
||||
11572705
|
Brady™
013617 |
Brown | 15.24 m x 254 mm |
£242.00
1 box |
Estimated Shipment: 08-08-2024
Log in to see stock. |
||||
11562695
|
Brady™
013606 |
Gold | 15.24 m x 152.4 mm |
£205.00
1 box |
Estimated Shipment: 06-09-2024
Log in to see stock. |
||||
11542655
|
Brady™
013558 |
Gold | 15.24 m x 203.20 mm |
£231.00
1 box |
Estimated Shipment: 06-09-2024
Log in to see stock. |
||||
11512645
|
Brady™
013545 |
Gold | 15.24 m x 101.6 mm |
£171.00
1 box |
Estimated Shipment: 06-09-2024
Log in to see stock. |
||||
11552705
|
Brady™
013615 |
Gold | 15.24 m x 254 mm |
£242.00
1 box |
Estimated Shipment: 06-09-2024
Log in to see stock. |
||||
11582635
|
Brady™
013542 |
Green | 15.24 m x 101.6 mm |
£171.00
1 box |
Estimated Shipment: 08-08-2024
Log in to see stock. |
||||
11532695
|
Brady™
013603 |
Green | 15.24 m x 152.4 mm |
£205.00
1 box |
Estimated Shipment: 08-08-2024
Log in to see stock. |
||||
11512655
|
Brady™
013555 |
Green | 15.24 m x 203.20 mm |
£231.00
1 box |
Estimated Shipment: 06-09-2024
Log in to see stock. |
||||
11522705
|
Brady™
013612 |
Green | 15.24 m x 254 mm |
£242.00
1 box |
Estimated Shipment: 08-08-2024
Log in to see stock. |
||||
11522645
|
Brady™
013546 |
Gray | 15.24 m x 101.6 mm |
£171.00
1 box |
Estimated Shipment: 06-09-2024
Log in to see stock. |
||||
11572695
|
Brady™
013607 |
Gray | 15.24 m x 152.4 mm |
£205.00
1 box |
Estimated Shipment: 06-09-2024
Log in to see stock. |
||||
11552655
|
Brady™
013559 |
Gray | 15.24 m x 203.20 mm |
£231.00
1 box |
Estimated Shipment: 06-09-2024
Log in to see stock. |
||||
11562705
|
Brady™
013616 |
Gray | 15.24 m x 254 mm |
£242.00
1 box |
Estimated Shipment: 08-08-2024
Log in to see stock. |
||||
11542695
|
Brady™
013604 |
Orange | 15.24 m x 152.4 mm |
£205.00
1 box |
Estimated Shipment: 06-09-2024
Log in to see stock. |
||||
11592635
|
Brady™
013543 |
Orange | 15.24 m x 101.6 mm |
£171.00
1 box |
Estimated Shipment: 06-09-2024
Log in to see stock. |
||||
11522655
|
Brady™
013556 |
Orange | 15.24 m x 203.20 mm |
£231.00
1 box |
Estimated Shipment: 06-09-2024
Log in to see stock. |
||||
11542645
|
Brady™
013548 |
Purple | 15.24 m x 101.6 mm |
£171.00
1 box |
Estimated Shipment: 06-09-2024
Log in to see stock. |
||||
11532705
|
Brady™
013613 |
Orange | 15.24 m x 254 mm |
£242.00
1 box |
Estimated Shipment: 08-08-2024
Log in to see stock. |
||||
11592695
|
Brady™
013609 |
Purple | 15.24 m x 152.4 mm |
£205.00
1 box |
Estimated Shipment: 06-09-2024
Log in to see stock. |
||||
11572655
|
Brady™
013561 |
Purple | 15.24 m x 203.20 mm |
£231.00
1 box |
Estimated Shipment: 06-09-2024
Log in to see stock. |
||||
11582705
|
Brady™
013618 |
Purple | 15.24 m x 254 mm |
£242.00
1 box |
Estimated Shipment: 08-08-2024
Log in to see stock. |
||||
11562635
|
Brady™
013540 |
Red | 15.24 m x 101.6 mm |
£171.00
1 box |
Estimated Shipment: 08-08-2024
Log in to see stock. |
||||
11512695
|
Brady™
013601 |
Red | 15.24 m x 152.4 mm |
£205.00
1 box |
Estimated Shipment: 08-08-2024
Log in to see stock. |
||||
11592645
|
Brady™
013553 |
Red | 15.24 m x 203.20 mm |
£231.00
1 box |
Estimated Shipment: 06-09-2024
Log in to see stock. |
||||
11502705
|
Brady™
013610 |
Red | 15.24 m x 254 mm |
£242.00
1 box |
Estimated Shipment: 08-08-2024
Log in to see stock. |
||||
11542635
|
Brady™
013538 |
White | 15.24 m x 101.6 mm |
£171.00
1 box |
Estimated Shipment: 08-08-2024
Log in to see stock. |
||||
11572645
|
Brady™
013551 |
White | 15.24 m x 203.20 mm |
£231.00
1 box |
Estimated Shipment: 08-08-2024
Log in to see stock. |
||||
11552645
|
Brady™
013549 |
White | 15.24 m x 152.4 mm |
£205.00
1 box |
Estimated Shipment: 08-08-2024
Log in to see stock. |
||||
11582655
|
Brady™
013562 |
White | 15.24 m x 254 mm |
£242.00
1 box |
Estimated Shipment: 08-08-2024
Log in to see stock. |
||||
11552635
|
Brady™
013539 |
Yellow | 15.24 m x 101.6 mm |
£171.00
1 box |
Estimated Shipment: 08-08-2024
Log in to see stock. |
||||
11562645
|
Brady™
013550 |
Yellow | 15.24 m x 152.4 mm |
£205.00
1 box |
Estimated Shipment: 08-08-2024
Log in to see stock. |
||||
11582645
|
Brady™
013552 |
Yellow | 15.24 m x 203.20 mm |
£231.00
1 box |
Estimated Shipment: 08-08-2024
Log in to see stock. |
||||
Description
- Brady's B-595 vinyl will adhere to difficult low surface energy items, including: PVC piping, blow-molded equipment cases, ABS plastics, powder-coated surfaces and equipment made from recycled plastics.
- The B-595 material also excels at sticking to highly textured surfaces such as painted cinder block walls, finished uneven wood, textured plastics, rough-pipes and paper-jacketed pipes.
Specifications
RoHS compliant to 2005/618/EC MCV amendment to RoHS Directive 2002/95/EC | |
Gloss | |
Vinyl | |
Labels | |
BBP85, PowerMark Industrial Sign and Label Printer | |
1 | |
1 Roll/Box | |
MarkWare | |
-40°C to +82°C | |
0.102 mm | |
15.24 m |
Permanent Acrylic, Permanent | |
Roll | |
Thermal Transfer | |
Component and equipment labelling, Electrical labelling, Facility Identification, Floor marking, Laboratory labelling, Lean and 5S labelling, Pipe and valve marking, Warehouse marking | |
60° 50 Gardner Units | |
-40°C | |
1 Ea. | |
-40°F to +180°F | |
0.004 in. | |
50 ft. |