missing translation for 'onlineSavingsMsg'
Learn More
Praseodymium powder, -200 mesh, 99.9% (REO), Thermo Scientific Chemicals Product Code.: 15452557

Praseodymium powder, -200 mesh, 99.9% (REO), Thermo Scientific Chemicals

Product Code. 15452557
25 g
Click to view available options
Quantity:
25 g
This item is not returnable. View return policy

Product Code. 15452557

Brand: Thermo Scientific Alfa Aesar 045910.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under Argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-10-0
Molecular Formula Pr
Molecular Weight (g/mol) 140.91
MDL Number MFCD00011174
InChI Key PUDIUYLPXJFUGB-UHFFFAOYSA-N
Synonym unii-nkn7eza750, nkn7eza750, ingot, praseodimio, praseodyme, praseodym, atom, chips, powder, acmc-1bktg
PubChem CID 23942
ChEBI CHEBI:49828
IUPAC Name praseodymium
SMILES [Pr]

Specifications

Color Silver
UN Number UN3089
Quantity 25 g
Percent Purity 99.9%
Mesh Size −200 Mesh
Odor Odorless
Sensitivity Air and moisture sensitive
Form Powder
Assay (REO)
Chemical Name or Material Praseodymium powder

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberU16K010 Certificate TypeCertificate of Analysis Date28/08/2024 Catalog Number045910.14,045910.22
Lot NumberT08K039 Certificate TypeCertificate of Analysis Date24/07/2024 Catalog Number045910.14,045910.22
Lot NumberR29K021 Certificate TypeCertificate of Analysis Date11/06/2024 Catalog Number045910.14,045910.22
Lot NumberP05K005 Certificate TypeCertificate of Analysis Date25/03/2024 Catalog Number045910.14,045910.22
Lot NumberS29H005 Certificate TypeCertificate of Analysis Date08/06/2022 Catalog Number045910.14,045909.06,045909.14

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Praseodymium powder, -200 mesh, 99.9% (REO), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.