missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brady™ Pre-cut Blank Labels for BBP3X, S3XXX, and i3300 Printers
The material of these labels is ideal for indoor and outdoor facility labels.
Brand: Brady™ 120935
Additional Details : Weight : 0.00050kg
Description
- Ideal for 5S labels, safety labels, warning labels, pipe markers, equipment labels and visual workplace labels
- Vinyl material
Specifications
Not Autoclavable | |
Yellow | |
Vinyl (B-7569) | |
Non-sterile | |
BBP™30, BBP™31, BBP™33, BBP™35 Multicolor, BBP™37 Multicolor, BradyPrinter i3300, S3000, S3100, and i5300 Industrial Label Printers | |
300 Labels | |
-20°C to +75°C | |
82.6 mm | |
75 mm |
Permanent Acrylic | |
Gloss | |
Thermal Transfer | |
Facility Identification, Hazard Communication Identification, Electrical Labeling | |
1 | |
Rectangle | |
0.102 mm | |
57 mm |